Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATP1B1 SIRNA Assays ›
          Gene Symbol:
          ATP1B1
          Gene Name:
          ATPase Na+/K+ transporting subunit beta 1
          Gene Aliases:
          ATP1B
          Chromosome Location:
          Chr.1: 169106709 - 169132722 on Build GRCh38
          UniGene ID:
          Hs.291196
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 10437

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ATP1B1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001677.3 NP_001668.1 6 792
          GenBank mRNA AK290181.1 - 6 805
          AK296059.1 - 6 680
          AK316043.1 - 6 737
          AY545986.1 - 6 671
          AY946016.1 - 2 121
          BC000006.2 - 6 757 3506311
          BG702024.1 - 2 78
          BT009787.1 - 6 671
          CD106723.1 - 2 116
          CR407616.1 - 6 671
          U16799.1 - 6 716

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          positive regulation of defense response to virus by host
          cellular calcium ion homeostasis
          cellular sodium ion homeostasis
          cell adhesion
          potassium ion import
          establishment or maintenance of transmembrane electrochemical gradient
          regulation of gene expression
          regulation of cardiac muscle contraction by calcium ion signaling
          cellular potassium ion homeostasis
          positive regulation of ATPase activity
          ion transmembrane transport
          sodium ion export from cell
          protein transport into plasma membrane raft
          ATP metabolic process
          protein stabilization
          leukocyte migration
          relaxation of cardiac muscle
          cardiac muscle contraction
          protein localization to plasma membrane
          membrane repolarization
          membrane repolarization during cardiac muscle cell action potential
          cell communication by electrical coupling involved in cardiac conduction
          ATP hydrolysis coupled transmembrane transport
          mitophagy in response to mitochondrial depolarization
          xenophagy
          positive regulation of potassium ion transmembrane transporter activity
          positive regulation of sodium ion export from cell
          positive regulation of calcium:sodium antiporter activity
          positive regulation of potassium ion import
          regulation of cardiac conduction
          potassium ion import across plasma membrane
          ATPase activator activity
          sodium:potassium-exchanging ATPase activity
          protein binding
          ATP binding
          protein C-terminus binding
          drug binding
          ATPase activity
          MHC class II protein complex binding
          potassium ion binding
          sodium ion binding
          ATPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0