Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KCNMA1 SIRNA Assays ›
Gene Symbol:
KCNMA1
Gene Name:
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases:
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
Chromosome Location:
Chr.10: 76869601 - 77637819 on Build GRCh38
UniGene ID:
Hs.144795
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
104416
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KCNMA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001014797.2
NP_001014797.1
14
1818
NM_001161352.1
NP_001154824.1
14
1818
NM_001161353.1
NP_001154825.1
14
1818
NM_001271518.1
NP_001258447.1
13
1656
NM_001271519.1
NP_001258448.1
14
1818
NM_001322829.1
NP_001309758.1
14
1818
NM_001322830.1
NP_001309759.1
14
1818
NM_001322832.1
NP_001309761.1
14
1818
NM_001322835.1
NP_001309764.1
14
1818
NM_001322836.1
NP_001309765.1
14
1818
NM_001322837.1
NP_001309766.1
14
1818
NM_001322838.1
NP_001309767.1
15
1778
NM_002247.3
NP_002238.2
14
1818
XM_005269776.3
XP_005269833.1
14
1807
XM_005269778.1
XP_005269835.1
14
1807
XM_005269781.2
XP_005269838.1
14
1739
XM_005269787.3
XP_005269844.1
14
1807
XM_005269789.2
XP_005269846.1
14
1739
XM_005269792.2
XP_005269849.1
14
1739
XM_005269796.2
XP_005269853.1
14
1739
XM_006717826.2
XP_006717889.1
14
1739
XM_011539773.1
XP_011538075.1
15
1939
XM_011539774.1
XP_011538076.1
15
1939
XM_011539775.1
XP_011538077.1
15
1939
XM_011539776.1
XP_011538078.1
15
1939
XM_011539777.1
XP_011538079.1
14
1807
XM_011539778.2
XP_011538080.1
14
1739
XM_011539779.1
XP_011538081.1
15
1939
XM_011539780.1
XP_011538082.1
14
1807
XM_011539781.2
XP_011538083.1
15
1939
XM_011539782.2
XP_011538084.1
15
1891
XM_011539783.1
XP_011538085.1
14
1807
XM_011539784.1
XP_011538086.1
15
1939
XM_011539785.1
XP_011538087.1
14
1807
XM_017016207.1
XP_016871696.1
15
1939
XM_017016208.1
XP_016871697.1
15
1939
XM_017016209.1
XP_016871698.1
15
1939
XM_017016210.1
XP_016871699.1
15
1939
XM_017016211.1
XP_016871700.1
15
1939
XM_017016212.1
XP_016871701.1
15
1939
XM_017016213.1
XP_016871702.1
15
1939
XM_017016214.1
XP_016871703.1
15
1939
XM_017016215.1
XP_016871704.1
14
1807
XM_017016216.1
XP_016871705.1
15
1939
XM_017016217.1
XP_016871706.1
15
1891
XM_017016218.1
XP_016871707.1
13
1264
XM_017016219.1
XP_016871708.1
15
1939
XM_017016222.1
XP_016871711.1
14
1807
XM_017016223.1
XP_016871712.1
12
1116
GenBank mRNA
AB113382.1
-
14
1583
AB113575.1
-
14
1569
AB209488.1
-
13
1224
AB524033.1
-
13
1479
AF025999.1
-
14
1569
AF118141.1
-
14
1446
AK128392.1
-
5
2074
AK310379.1
-
15
1778
AY040849.1
-
14
1583
BC137115.1
-
14
1721
9021112
BC137137.1
-
14
1721
9021134
BC144496.1
-
14
1721
9053023
BX648925.1
-
9
800
U02632.1
-
7
676
U09384.1
-
14
1636
U11058.2
-
14
1532
U11717.1
-
14
1593
U13913.1
-
14
1719
U23767.1
-
14
1641
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
potassium ion transport
response to osmotic stress
cellular potassium ion homeostasis
response to carbon monoxide
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of apoptotic process
negative regulation of cell volume
response to calcium ion
micturition
smooth muscle contraction involved in micturition
potassium ion transmembrane transport
actin binding
voltage-gated potassium channel activity
protein binding
calcium-activated potassium channel activity
metal ion binding
large conductance calcium-activated potassium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0