Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MTMR2 SIRNA Assays ›
          Gene Symbol:
          MTMR2
          Gene Name:
          myotubularin related protein 2
          Gene Aliases:
          CMT4B, CMT4B1
          Chromosome Location:
          Chr.11: 95832880 - 95924207 on Build GRCh38
          UniGene ID:
          Hs.181326
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 104912
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MTMR2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243571.1 NP_001230500.1 7 835
          NM_016156.5 NP_057240.3 4 568
          NM_201278.2 NP_958435.1 6 762
          NM_201281.2 NP_958438.1 5 639
          XM_005274374.2 XP_005274431.1 5 480
          XM_005274375.2 XP_005274432.1 5 594
          XM_006718934.2 XP_006718997.1 6 674
          XM_006718935.2 XP_006718998.1 6 604
          XM_006718936.3 XP_006718999.1 5 606
          XM_011543058.2 XP_011541360.1 7 696
          XM_011543059.2 XP_011541361.1 7 530
          XM_017018517.1 XP_016874006.1 6 701
          XM_017018518.1 XP_016874007.1 5 483
          GenBank mRNA AB028996.1 - 4 656
          AK291277.1 - 4 342
          AK293243.1 - 6 473
          AK302940.1 - 5 365
          AL705005.1 - 6 613
          BC034474.1 - 6 659
          BC040432.1 - 4 320
          BC052990.1 - 4 453 6501680
          BM475570.1 - 4 453
          BP358035.1 - 4 568
          BX647997.1 - 5 533

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00269534_m1

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of receptor internalization
          protein dephosphorylation
          phosphatidylinositol biosynthetic process
          negative regulation of myelination
          myelin assembly
          peptidyl-tyrosine dephosphorylation
          negative regulation of endocytosis
          inositol phosphate dephosphorylation
          phosphatidylinositol dephosphorylation
          neuron development
          protein tetramerization
          negative regulation of excitatory postsynaptic potential
          dendritic spine maintenance
          positive regulation of early endosome to late endosome transport
          negative regulation of receptor catabolic process
          phosphatidylinositol-3-phosphatase activity
          protein tyrosine phosphatase activity
          protein binding
          protein tyrosine/serine/threonine phosphatase activity
          protein homodimerization activity
          phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0