Search
Search

HTR3A| Gene Symbol: HTR3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000869.5 | NP_000860.2 | 2 | 430 | ||
| NM_001161772.2 | NP_001155244.1 | 2 | 189 | |||
| NM_213621.3 | NP_998786.2 | 2 | 430 | |||
| NR_046363.1 | 2 | 430 | ||||
| GenBank mRNA | AF498984.1 | - | 2 | 179 | ||
| AJ003078.1 | - | 2 | 219 | |||
| AJ003079.1 | - | 2 | 219 | |||
| AJ003080.1 | - | 2 | 219 | |||
| AK299973.1 | - | 2 | 189 | |||
| AK304630.1 | - | 2 | 432 | |||
| AK313896.1 | - | 2 | 309 | |||
| BC002354.2 | - | 2 | 246 | 2821710 | ||
| BC004453.1 | - | 2 | 298 | 2819400 | ||
| BT007204.1 | - | 2 | 197 | |||
| CD014118.1 | - | 2 | 317 | |||
| D49394.1 | - | 2 | 398 | |||
| S82612.1 | - | 2 | 221 | |||