Search
Search

CPA3| Gene Symbol: CPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001870.3 | NP_001861.2 | 2 | 156 | ||
| GenBank mRNA | AK313628.1 | - | 2 | 116 | ||
| BC012613.1 | - | 2 | 119 | 4104766 | ||
| BG501260.1 | - | 2 | 158 | |||
| DR005288.1 | - | 2 | 106 | |||
| M27717.1 | - | 2 | 104 | |||