Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PSMA5 SIRNA Assays ›
          Gene Symbol:
          PSMA5
          Gene Name:
          proteasome subunit alpha 5
          Gene Aliases:
          PSC5, ZETA
          Chromosome Location:
          Chr.1: 109399031 - 109426486 on Build GRCh38
          UniGene ID:
          Hs.485246
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 105609

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PSMA5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199772.1 NP_001186701.1 2 247
          NM_001199773.1 NP_001186702.1 3 222
          NM_001199774.1 NP_001186703.1 3 234
          NM_002790.3 NP_002781.2 3 314
          GenBank mRNA AK225990.1 - 3 302
          AK304448.1 - 2 239
          AK313351.1 - 3 314
          BC102018.1 - 3 266 40030391
          BC102019.1 - 3 266 40030392
          BC102020.1 - 3 266 40030394
          BC103751.1 - 3 289 6013212
          BG743267.1 - 3 300
          BI825400.1 - 3 222
          BM793388.1 - 3 186
          BT019717.1 - 3 196
          CR456847.1 - 3 196
          DC320780.1 - 3 221
          DC324021.1 - 3 234
          DC324498.1 - 2 247
          GU727639.1 - 3 196
          X61970.1 - 3 217

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          protein polyubiquitination
          stimulatory C-type lectin receptor signaling pathway
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          regulation of cellular amino acid metabolic process
          anaphase-promoting complex-dependent catabolic process
          tumor necrosis factor-mediated signaling pathway
          NIK/NF-kappaB signaling
          Fc-epsilon receptor signaling pathway
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of mRNA stability
          T cell receptor signaling pathway
          negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
          Wnt signaling pathway, planar cell polarity pathway
          negative regulation of canonical Wnt signaling pathway
          positive regulation of canonical Wnt signaling pathway
          threonine-type endopeptidase activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0