Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CFLAR SIRNA Assays ›
Gene Symbol:
CFLAR
Gene Name:
CASP8 and FADD like apoptosis regulator
Gene Aliases:
CASH, CASP8AP1, CLARP, Casper, FLAME, FLAME-1, FLAME1, FLIP, I-FLICE, MRIT, c-FLIP, c-FLIPL, c-FLIPR, c-FLIPS
Chromosome Location:
Chr.2: 201116015 - 201172688 on Build GRCh38
UniGene ID:
Hs.390736
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
105636
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CFLAR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127183.2
NP_001120655.1
2
683
NM_001127184.2
NP_001120656.1
2
655
NM_001202516.1
NP_001189445.1
2
655
NM_001308042.1
NP_001294971.1
1
327
NM_001308043.1
NP_001294972.1
2
705
NM_003879.5
NP_003870.4
2
655
XM_017005189.1
XP_016860678.1
2
646
XM_017005190.1
XP_016860679.1
2
519
XM_017005191.1
XP_016860680.1
2
598
XM_017005192.1
XP_016860681.1
2
646
XM_017005193.1
XP_016860682.1
2
2663
XM_017005194.1
XP_016860683.1
2
519
XM_017005195.1
XP_016860684.1
2
636
XM_017005197.1
XP_016860686.1
2
2663
XM_017005198.1
XP_016860687.1
2
519
GenBank mRNA
AB209600.1
-
2
402
AF005774.1
-
2
625
AF005775.1
-
1
190
AF009616.1
-
2
602
AF009617.1
-
1
190
AF009618.1
-
1
190
AF009619.1
-
1
190
AF010127.1
-
2
600
AF015450.1
-
2
694
AF015451.1
-
1
190
AF015452.1
-
1
190
AF041458.1
-
1
205
AF041460.1
-
1
190
AF041461.1
-
1
145
AK289913.1
-
2
479
AK308767.1
-
2
565
AK315208.1
-
2
426
BC001602.1
-
2
449
3542993
BT006751.1
-
1
190
BX457155.2
-
2
672
U85059.1
-
1
190
U97074.1
-
2
572
U97075.1
-
1
483
Y14039.1
-
2
671
Y14040.1
-
2
671
Back To Top
More Information
Panther Classification:
Molecular Function -
protease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
apoptotic process
skeletal muscle tissue development
skeletal muscle atrophy
regulation of skeletal muscle satellite cell proliferation
skeletal myofibril assembly
viral process
negative regulation of apoptotic process
positive regulation of catalytic activity
positive regulation of I-kappaB kinase/NF-kappaB signaling
skeletal muscle tissue regeneration
positive regulation of NF-kappaB transcription factor activity
regulation of necroptotic process
execution phase of apoptosis
negative regulation of myoblast fusion
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway
protease binding
death receptor binding
protein binding
enzyme activator activity
protein complex binding
cysteine-type endopeptidase activity involved in execution phase of apoptosis
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0