Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RB1 SIRNA Assays ›
          Gene Symbol:
          RB1
          Gene Name:
          RB transcriptional corepressor 1
          Gene Aliases:
          OSRC, PPP1R130, RB, p105-Rb, pRb, pp110
          Chromosome Location:
          Chr.13: 48303747 - 48481890 on Build GRCh38
          UniGene ID:
          Hs.408528
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 106040

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000321.2 NP_000312.2 19 2023
          XM_011535171.2 XP_011533473.1 18 1612
          GenBank mRNA AB208788.1 - 19 1954
          AK291258.1 - 19 2044
          AK299179.1 - 20 1934
          AK300284.1 - 20 1960
          AK307730.1 - 19 2011
          BC039060.1 - 19 2009 5142020
          BC040540.1 - 19 2019 5267622
          L41870.1 - 19 1994
          M15400.1 - 19 1995
          M28419.1 - 19 1995
          M33647.1 - 19 1860

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle checkpoint
          G1/S transition of mitotic cell cycle
          regulation of transcription involved in G1/S transition of mitotic cell cycle
          chromatin remodeling
          transcription, DNA-templated
          negative regulation of protein kinase activity
          cell cycle arrest
          negative regulation of transcription from RNA polymerase II promoter during mitosis
          mitotic cell cycle checkpoint
          Ras protein signal transduction
          regulation of mitotic cell cycle
          negative regulation of gene expression
          viral process
          covalent chromatin modification
          androgen receptor signaling pathway
          sister chromatid biorientation
          neuron projection development
          maintenance of mitotic sister chromatid cohesion
          glial cell apoptotic process
          skeletal muscle cell differentiation
          neuron maturation
          enucleate erythrocyte differentiation
          negative regulation of sequence-specific DNA binding transcription factor activity
          regulation of lipid kinase activity
          myoblast differentiation
          positive regulation of macrophage differentiation
          positive regulation of mitotic metaphase/anaphase transition
          negative regulation of smoothened signaling pathway
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          digestive tract development
          cell morphogenesis involved in neuron differentiation
          negative regulation of epithelial cell proliferation
          striated muscle cell differentiation
          cell division
          neuron apoptotic process
          protein localization to chromosome, centromeric region
          cellular response to xenobiotic stimulus
          regulation of cohesin loading
          negative regulation of transcription involved in G1/S transition of mitotic cell cycle
          regulation of centromere complex assembly
          hepatocyte apoptotic process
          negative regulation of G1/S transition of mitotic cell cycle
          positive regulation of transcription regulatory region DNA binding
          core promoter binding
          RNA polymerase II activating transcription factor binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          protein binding
          transcription factor binding
          kinase binding
          ubiquitin protein ligase binding
          identical protein binding
          androgen receptor binding
          phosphoprotein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0