Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HTR2B SIRNA Assays ›
          Gene Symbol:
          HTR2B
          Gene Name:
          5-hydroxytryptamine receptor 2B
          Gene Aliases:
          5-HT(2B), 5-HT-2B, 5-HT2B
          Chromosome Location:
          Chr.2: 231108230 - 231125778 on Build GRCh38
          UniGene ID:
          Hs.421649
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 106295

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HTR2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000867.4 NP_000858.3 3 999
          NM_001320758.1 NP_001307687.1 3 616
          XM_005246520.3 XP_005246577.1 2 949
          XM_006712482.2 XP_006712545.1 3 1196
          GenBank mRNA AK313741.1 - 3 931
          AY136751.1 - 2 494
          BC063123.1 - 3 1003 30321290
          X77307.1 - 2 549
          Z36748.1 - 2 540

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          neural crest cell migration
          positive regulation of cytokine production
          positive regulation of endothelial cell proliferation
          G-protein coupled receptor internalization
          heart morphogenesis
          cardiac muscle hypertrophy
          cGMP biosynthetic process
          cellular calcium ion homeostasis
          G-protein coupled receptor signaling pathway
          activation of phospholipase C activity
          protein kinase C-activating G-protein coupled receptor signaling pathway
          serotonin receptor signaling pathway
          behavior
          positive regulation of cell proliferation
          negative regulation of autophagy
          positive regulation of phosphatidylinositol biosynthetic process
          neural crest cell differentiation
          phosphatidylinositol 3-kinase signaling
          intestine smooth muscle contraction
          phosphorylation
          calcium-mediated signaling
          hepatic stellate cell activation
          vasoconstriction
          response to drug
          negative regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of MAP kinase activity
          positive regulation of GTPase activity
          inositol phosphate metabolic process
          embryonic morphogenesis
          positive regulation of cytokine secretion
          regulation of behavior
          positive regulation of nitric-oxide synthase activity
          release of sequestered calcium ion into cytosol
          positive regulation of cell division
          negative regulation of cell death
          ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          protein kinase C signaling
          cellular response to amine stimulus
          cellular response to temperature stimulus
          cellular response to serotonin
          G-protein alpha-subunit binding
          G-protein coupled serotonin receptor activity
          GTPase activator activity
          drug binding
          serotonin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gl4ql:80/100.66.76.174:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0