Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ARNTL SIRNA Assays ›
          Gene Symbol:
          ARNTL
          Gene Name:
          aryl hydrocarbon receptor nuclear translocator like
          Gene Aliases:
          BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5
          Chromosome Location:
          Chr.11: 13276613 - 13387268 on Build GRCh38
          UniGene ID:
          Hs.65734
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 106398

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ARNTL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001030272.2 NP_001025443.1 18 2039
          NM_001030273.2 NP_001025444.1 19 2110
          NM_001178.5 NP_001169.3 19 2094
          NM_001297719.1 NP_001284648.1 19 2097
          NM_001297722.1 NP_001284651.1 18 2042
          NM_001297724.1 NP_001284653.1 19 2106
          XM_006718233.3 XP_006718296.1 18 1999
          XM_006718234.1 XP_006718297.1 19 2065
          XM_011520105.2 XP_011518407.2 17 2160
          XM_011520107.2 XP_011518409.2 17 2140
          XM_011520108.1 XP_011518410.1 19 2105
          XM_011520109.1 XP_011518411.1 17 1860
          XM_011520110.1 XP_011518412.1 20 2120
          XM_011520111.1 XP_011518413.1 18 1935
          XM_011520112.2 XP_011518414.1 20 2105
          XM_011520113.1 XP_011518415.1 20 2102
          XM_017017738.1 XP_016873227.1 17 2161
          XM_017017739.1 XP_016873228.1 17 2142
          XM_017017740.1 XP_016873229.1 18 1996
          XM_017017741.1 XP_016873230.1 17 1857
          XM_017017742.1 XP_016873231.1 17 1845
          XM_017017743.1 XP_016873232.1 17 1842
          XM_017017744.1 XP_016873233.1 20 2117
          XM_017017745.1 XP_016873234.1 19 2062
          XM_017017746.1 XP_016873235.1 19 2060
          XM_017017747.1 XP_016873236.1 18 1920
          XM_017017748.1 XP_016873237.1 18 1974
          GenBank mRNA AB000812.1 - 16 1678
          AB000814.1 - 5 494
          AF044288.1 - 19 2007
          AK095749.1 - 19 1995
          AK128317.1 - 4 3112
          AK291510.1 - 18 1955
          AK315174.1 - 19 2035
          BC016674.1 - 16 1641 3921870
          BC031214.1 - 19 1938 4793007
          BC041129.1 - 19 2018 5753529
          D89722.1 - 16 1654
          U51627.1 - 13 1636
          U60415.1 - 18 1941

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_024362, XM_006230030, XM_006230029, XM_008759716, AF015953, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001243048, NM_007489, AK146628, AB015203, AK147145, and others.

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus, translocation
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          spermatogenesis
          circadian rhythm
          negative regulation of TOR signaling
          circadian regulation of gene expression
          regulation of protein catabolic process
          regulation of hair cycle
          positive regulation of circadian rhythm
          proteasome-mediated ubiquitin-dependent protein catabolic process
          negative regulation of fat cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of neurogenesis
          regulation of insulin secretion
          regulation of cell cycle
          response to redox state
          maternal process involved in parturition
          positive regulation of canonical Wnt signaling pathway
          oxidative stress-induced premature senescence
          regulation of type B pancreatic cell development
          negative regulation of glucocorticoid receptor signaling pathway
          regulation of cellular senescence
          positive regulation of skeletal muscle cell differentiation
          transcription regulatory region sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          core promoter sequence-specific DNA binding
          core promoter binding
          transcriptional activator activity, RNA polymerase II transcription factor binding
          DNA binding
          protein binding
          aryl hydrocarbon receptor binding
          bHLH transcription factor binding
          sequence-specific DNA binding
          protein heterodimerization activity
          Hsp90 protein binding
          repressing transcription factor binding
          E-box binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0