Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SMARCA4 SIRNA Assays ›
Gene Symbol:
SMARCA4
Gene Name:
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4
Gene Aliases:
BAF190, BAF190A, BRG1, CSS4, MRD16, RTPS2, SNF2, SNF2L4, SNF2LB, SWI2, hSNF2b
Chromosome Location:
Chr.19: 10960922 - 11062282 on Build GRCh38
UniGene ID:
Hs.327527
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
106960
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SMARCA4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128844.1
NP_001122316.1
13
2222
NM_001128845.1
NP_001122317.1
11
1862
NM_001128846.1
NP_001122318.1
11
1862
NM_001128847.1
NP_001122319.1
11
1862
NM_001128848.1
NP_001122320.1
11
1862
NM_001128849.1
NP_001122321.1
12
1936
NM_003072.3
NP_003063.2
12
2146
XM_006722845.2
XP_006722908.1
12
2068
XM_006722846.2
XP_006722909.1
12
2162
XM_011528198.1
XP_011526500.1
13
2143
XM_017027160.1
XP_016882649.1
13
2143
XM_017027161.1
XP_016882650.1
13
2143
XM_017027162.1
XP_016882651.1
13
2143
XM_017027163.1
XP_016882652.1
13
2143
XM_017027164.1
XP_016882653.1
13
2143
XM_017027165.1
XP_016882654.1
13
2143
XM_017027166.1
XP_016882655.1
13
2143
XM_017027167.1
XP_016882656.1
13
2143
XM_017027168.1
XP_016882657.1
13
2143
GenBank mRNA
AB209313.2
-
11
1736
AK290033.1
-
13
2143
BC136644.1
-
10
2082
9020634
BC150298.1
-
12
2067
8860365
D26156.1
-
12
1919
EU430756.1
-
11
1862
EU430757.1
-
11
1862
EU430758.1
-
11
1862
EU430759.1
-
11
1862
U29175.1
-
12
1940
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011417, NM_001174078, NM_001174079, AK135308, AK147285, and others.
Panther Classification:
Molecular Function -
DNA helicase
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
neural retina development
nucleosome disassembly
chromatin remodeling
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
negative regulation of transcription from RNA polymerase II promoter during mitosis
spermatid development
nervous system development
covalent chromatin modification
positive regulation of Wnt signaling pathway
negative regulation of cell growth
ATP-dependent chromatin remodeling
positive regulation by host of viral transcription
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of sequence-specific DNA binding transcription factor activity
negative regulation of androgen receptor signaling pathway
positive regulation of transcription of nuclear large rRNA transcript from RNA polymerase I promoter
positive regulation of glucose mediated signaling pathway
positive regulation of pri-miRNA transcription from RNA polymerase II promoter
beta-catenin-TCF complex assembly
negative regulation of G1/S transition of mitotic cell cycle
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II transcription coactivator activity
RNA polymerase I CORE element sequence-specific DNA binding
p53 binding
transcription coactivator activity
transcription corepressor activity
helicase activity
protein binding
ATP binding
DNA-dependent ATPase activity
transcription factor binding
Tat protein binding
nucleosomal DNA binding
protein N-terminus binding
androgen receptor binding
DNA polymerase binding
lysine-acetylated histone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0