Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZMYM2 SIRNA Assays ›
          Gene Symbol:
          ZMYM2
          Gene Name:
          zinc finger MYM-type containing 2
          Gene Aliases:
          FIM, MYM, RAMP, SCLL, ZNF198
          Chromosome Location:
          Chr.13: 19957412 - 20091845 on Build GRCh38
          UniGene ID:
          Hs.507433
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 107145

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZMYM2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001190964.2 NP_001177893.1 10, 11 2049
          NM_001190965.2 NP_001177894.1 9, 10 2064
          NM_003453.4 NP_003444.1 10, 11 2157
          NM_197968.3 NP_932072.1 9, 10 2110
          XM_005266520.3 XP_005266577.1 9, 10 2069
          XM_011535216.2 XP_011533518.1 10, 11 1739
          XM_011535222.1 XP_011533524.1 9, 10 2000
          XM_011535223.2 XP_011533525.1 3, 4 4454
          XM_011535224.2 XP_011533526.1 3, 4 355
          XM_017020728.1 XP_016876217.1 9, 10 2000
          XM_017020729.1 XP_016876218.1 9, 10 1975
          XM_017020731.1 XP_016876220.1 10, 11 1792
          XM_017020732.1 XP_016876221.1 10, 11 1814
          XM_017020733.1 XP_016876222.1 11, 12 1841
          XM_017020734.1 XP_016876223.1 11, 12 1827
          XM_017020735.1 XP_016876224.1 10, 11 1791
          XM_017020736.1 XP_016876225.1 10, 11 1963
          XM_017020737.1 XP_016876226.1 10, 11 1738
          XM_017020738.1 XP_016876227.1 9, 10 2053
          XM_017020739.1 XP_016876228.1 10, 11 2080
          GenBank mRNA AF012126.1 - 3, 4 452
          AF035374.1 - 7, 8 1355
          AF060181.1 - 7, 8 1407
          AJ224901.1 - 10, 11 2043
          AK226118.1 - 9, 10 2044
          AK289741.1 - 9, 10 2035
          AK302917.1 - 4, 5 424
          BC036372.1 - 9, 10 2110 4823008
          BX647944.1 - 9, 10 2064
          BX648905.1 - 2, 3 2494
          Y13472.1 - 10, 11 2055

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          cytoskeleton organization
          multicellular organism development
          biological_process
          peptidyl-tyrosine phosphorylation
          regulation of cell morphogenesis
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          DNA binding
          protein tyrosine kinase activity
          zinc ion binding
          ubiquitin conjugating enzyme binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0