Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPFIA2 SIRNA Assays ›
          Gene Symbol:
          PPFIA2
          Gene Name:
          PTPRF interacting protein alpha 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 81257975 - 81759748 on Build GRCh38
          UniGene ID:
          Hs.506216
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 107194

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPFIA2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001220475.2 NP_001207404.1 30 3792
          NM_001220476.2 NP_001207405.1 31 3837
          NM_001220477.2 NP_001207406.1 28 3781
          NM_001220478.2 NP_001207407.1 28 3769
          NM_001220480.2 NP_001207409.1 21 2444
          NM_001282536.1 NP_001269465.1 29 3550
          NM_003625.4 NP_003616.2 32 3906
          XM_006719670.3 XP_006719733.1 30 11107
          XM_011538906.2 XP_011537208.1 28 3872
          XM_011538907.2 XP_011537209.1 30 3965
          XM_017020079.1 XP_016875568.1 33 3899
          XM_017020080.1 XP_016875569.1 31 4037
          XM_017020081.1 XP_016875570.1 32 4028
          XM_017020082.1 XP_016875571.1 32 3872
          XM_017020083.1 XP_016875572.1 31 4016
          XM_017020085.1 XP_016875574.1 30 4007
          XM_017020088.1 XP_016875577.1 32 4279
          XM_017020089.1 XP_016875578.1 31 3992
          XM_017020092.1 XP_016875581.1 31 3983
          XM_017020094.1 XP_016875583.1 32 3824
          XM_017020095.1 XP_016875584.1 31 3965
          XM_017020096.1 XP_016875585.1 30 3962
          XM_017020097.1 XP_016875586.1 31 3809
          XM_017020098.1 XP_016875587.1 31 3806
          XM_017020101.1 XP_016875590.1 30 3797
          XM_017020102.1 XP_016875591.1 30 3931
          XM_017020104.1 XP_016875593.1 30 3935
          XM_017020106.1 XP_016875595.1 29 3926
          XM_017020110.1 XP_016875599.1 30 4195
          XM_017020112.1 XP_016875601.1 29 3862
          XM_017020113.1 XP_016875602.1 29 3844
          XM_017020117.1 XP_016875606.1 29 3606
          XM_017020118.1 XP_016875607.1 31 3846
          XM_017020119.1 XP_016875608.1 28 3799
          XM_017020120.1 XP_016875609.1 30 3559
          GenBank mRNA AF034799.1 - 32 3914
          AK123372.1 - 21 2379
          AK294505.1 - 28 3579
          AK299853.1 - 28 3730
          AK302658.1 - 29 3521
          BC104912.1 - 30 3776 8143915
          BC143485.1 - 31 3821 9051993
          BM689808.1 - 4 420

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001108745, XM_008765336, XM_008765332, XM_008765331, XM_008765329, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001205341, NM_177373, BC065133, AK087448, EU568870, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          cell-matrix adhesion
          neurotransmitter secretion
          glutamate secretion
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline