Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIA1 SIRNA Assays ›
Gene Symbol:
PPFIA1
Gene Name:
PTPRF interacting protein alpha 1
Gene Aliases:
LIP.1, LIP1, LIPRIN
Chromosome Location:
Chr.11: 70270687 - 70384501 on Build GRCh38
UniGene ID:
Hs.530749
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
107197
Validated
Purification
In-Vivo Ready
Standard
HPLC
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
417.00
Your Price
Online offer:
417.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003626.3
NP_003617.1
23
3300
NM_177423.2
NP_803172.1
23
3300
NR_045286.1
24
3311
XM_006718716.3
XP_006718779.1
28
3625
XM_011545306.2
XP_011543608.1
28
3497
XM_011545307.2
XP_011543609.1
27
3598
XM_011545308.2
XP_011543610.1
27
3595
XM_011545309.2
XP_011543611.1
28
3625
XM_011545310.2
XP_011543612.1
27
3550
XM_011545311.2
XP_011543613.1
26
3538
XM_011545312.2
XP_011543614.1
26
3535
XM_011545313.2
XP_011543615.1
27
3508
XM_011545314.2
XP_011543616.1
26
3478
XM_011545315.2
XP_011543617.1
26
3433
XM_011545316.2
XP_011543618.1
25
3418
XM_011545317.2
XP_011543619.1
24
3388
XM_011545318.2
XP_011543620.1
13
1730
XM_011545319.1
XP_011543621.1
25
3406
XM_017018448.1
XP_016873937.1
25
3505
XM_017018449.1
XP_016873938.1
24
3343
GenBank mRNA
AB209314.1
-
24
3238
AK123331.1
-
1877
AK124621.1
-
3
1845
BC034046.1
-
23
3263
4794300
BX648037.1
-
22
2600
D49354.1
-
8
1072
U22815.1
-
23
3314
U22816.1
-
23
3314
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00170349_m1
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
signal transduction
neurotransmitter secretion
glutamate secretion
negative regulation of stress fiber assembly
negative regulation of establishment of protein localization to plasma membrane
signal transducer activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline