Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CADPS SIRNA Assays ›
Gene Symbol:
CADPS
Gene Name:
calcium dependent secretion activator
Gene Aliases:
CADPS1, CAPS, CAPS1, UNC-31
Chromosome Location:
Chr.3: 62398346 - 62875389 on Build GRCh38
UniGene ID:
Hs.654933
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
107217
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CADPS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003716.3
NP_003707.2
6
1593
NM_183393.2
NP_899630.1
6
1593
NM_183394.2
NP_899631.1
6
1593
XM_006713378.2
XP_006713441.2
6
1588
XM_011534177.1
XP_011532479.1
6
1588
XM_011534178.1
XP_011532480.1
6
1588
XM_011534180.1
XP_011532482.1
6
1588
XM_011534182.1
XP_011532484.1
6
1588
XM_011534183.1
XP_011532485.1
6
1588
XM_011534189.1
XP_011532491.1
6
1588
XM_011534191.2
XP_011532493.1
6
1233
XM_011534192.1
XP_011532494.1
6
1588
XM_011534193.1
XP_011532495.1
6
1588
XM_011534194.2
XP_011532496.1
6
1233
XM_011534195.1
XP_011532497.1
6
1588
XM_011534196.1
XP_011532498.1
6
1588
XM_011534197.2
XP_011532499.1
6
1546
XM_011534198.2
XP_011532500.1
6
1233
XM_011534199.2
XP_011532501.1
6
1233
XM_011534200.2
XP_011532502.1
6
1233
XM_011534202.2
XP_011532504.1
6
1588
XM_011534203.1
XP_011532505.1
6
1588
XM_017007358.1
XP_016862847.1
6
1588
XM_017007359.1
XP_016862848.1
6
1588
XM_017007360.1
XP_016862849.1
6
1588
XM_017007361.1
XP_016862850.1
6
1588
XM_017007362.1
XP_016862851.1
6
1588
XM_017007363.1
XP_016862852.1
6
1588
XM_017007364.1
XP_016862853.1
6
1588
XM_017007365.1
XP_016862854.1
6
1588
XM_017007366.1
XP_016862855.1
6
1588
XM_017007367.1
XP_016862856.1
6
1233
XM_017007368.1
XP_016862857.1
6
1233
XM_017007369.1
XP_016862858.1
6
1588
XM_017007370.1
XP_016862859.1
6
1588
XM_017007371.1
XP_016862860.1
6
1588
XM_017007372.1
XP_016862861.1
6
1233
XM_017007373.1
XP_016862862.1
6
1588
XM_017007374.1
XP_016862863.1
6
1588
XM_017007375.1
XP_016862864.1
6
1233
XM_017007376.1
XP_016862865.1
6
1588
XM_017007377.1
XP_016862866.1
6
1233
XM_017007378.1
XP_016862867.1
6
1233
XM_017007379.1
XP_016862868.1
6
1588
XM_017007380.1
XP_016862869.1
6
1588
XM_017007381.1
XP_016862870.1
6
1588
XM_017007382.1
XP_016862871.1
6
1233
XM_017007383.1
XP_016862872.1
6
1588
XM_017007384.1
XP_016862873.1
6
1588
XM_017007385.1
XP_016862874.1
6
1588
XM_017007386.1
XP_016862875.1
6
1588
GenBank mRNA
AB032947.2
-
6
1583
AB593079.1
-
6
1546
AF458662.1
-
6
1543
AL833895.1
-
6
890
BC029460.1
-
2
276
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
protein transport
vesicle organization
synaptic vesicle priming
catecholamine secretion
dense core granule exocytosis
protein binding
lipid binding
protein kinase binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0