Search
Search

EBAG9| Gene Symbol: EBAG9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278938.1 | NP_001265867.1 | 4 | 349 | ||
| NM_004215.4 | NP_004206.1 | 4 | 636 | |||
| NM_198120.2 | NP_936056.1 | 4 | 469 | |||
| XM_017013960.1 | XP_016869449.1 | 4 | 832 | |||
| GenBank mRNA | AB007619.1 | - | 4 | 517 | ||
| AF006265.1 | - | 4 | 467 | |||
| AK290651.1 | - | 4 | 458 | |||
| AK308950.1 | - | 4 | 527 | |||
| AL515533.3 | - | 4 | 527 | |||
| AY515724.1 | - | 4 | 427 | |||
| AY653072.1 | - | 3 | 234 | |||
| BC005249.1 | - | 4 | 547 | 3930583 | ||
| BC017729.1 | - | 4 | 420 | 3453385 | ||
| BC022506.1 | - | 4 | 534 | 4815654 | ||
| BG616704.1 | - | 4 | 350 | |||
| BG707115.1 | - | 4 | 470 | |||
| BI194881.1 | - | 4 | 636 | |||
| CD108096.1 | - | 4 | 580 | |||
| CR456984.1 | - | 3 | 234 | |||