Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LRP8 SIRNA Assays ›
          Gene Symbol:
          LRP8
          Gene Name:
          LDL receptor related protein 8
          Gene Aliases:
          APOER2, HSZ75190, LRP-8, MCI1
          Chromosome Location:
          Chr.1: 53242364 - 53328149 on Build GRCh38
          UniGene ID:
          Hs.280387
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 107482

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LRP8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001018054.2 NP_001018064.1 8 1427
          NM_004631.4 NP_004622.2 8 1427
          NM_017522.4 NP_059992.3 7 1040
          NM_033300.3 NP_150643.2 6 917
          XM_005271173.3 XP_005271230.1 9 1376
          XM_005271174.3 XP_005271231.1 8 984
          XM_005271175.3 XP_005271232.1 7 945
          XM_006710881.3 XP_006710944.1 9 1376
          XM_006710882.3 XP_006710945.1 9 1376
          XM_011542095.2 XP_011540397.1 7 855
          XM_011542096.2 XP_011540398.1 9 1376
          XM_017002266.1 XP_016857755.1 9 1376
          XM_017002267.1 XP_016857756.1 7 821
          XM_017002268.1 XP_016857757.1 8 860
          GenBank mRNA AL119002.1 - 1 79
          BC051836.1 - 8 1332 6143442
          BE543079.1 - 4 256
          D50678.1 - 8 1292
          Z75190.1 - 7 803

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          retinoid metabolic process
          proteolysis
          lipid metabolic process
          endocytosis
          receptor-mediated endocytosis
          signal transduction
          cytokine-mediated signaling pathway
          ammon gyrus development
          cerebral cortex development
          positive regulation of CREB transcription factor activity
          reelin-mediated signaling pathway
          response to drug
          positive regulation of peptidyl-tyrosine phosphorylation
          modulation of synaptic transmission
          positive regulation of dendritic spine morphogenesis
          positive regulation of protein tyrosine kinase activity
          cellular response to growth factor stimulus
          cellular response to cholesterol
          positive regulation of dendrite development
          transmembrane signaling receptor activity
          low-density lipoprotein receptor activity
          calcium ion binding
          protein binding
          high-density lipoprotein particle binding
          kinesin binding
          very-low-density lipoprotein particle receptor activity
          apolipoprotein binding
          reelin receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0