Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LRRFIP2 SIRNA Assays ›
          Gene Symbol:
          LRRFIP2
          Gene Name:
          LRR binding FLII interacting protein 2
          Gene Aliases:
          HUFI-2
          Chromosome Location:
          Chr.3: 37052626 - 37176817 on Build GRCh38
          UniGene ID:
          Hs.740522
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 108003

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LRRFIP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134369.2 NP_001127841.1 13 1466
          NM_001282691.1 NP_001269620.1 17 1455
          NM_006309.3 NP_006300.1 26 2056
          NM_017724.2 NP_060194.1 12 1394
          XM_005265539.2 XP_005265596.1 27 3056
          XM_005265540.1 XP_005265597.1 28 2339
          XM_005265550.1 XP_005265607.1 19 1892
          XM_005265551.1 XP_005265608.1 18 1781
          XM_005265553.1 XP_005265610.1 15 1628
          XM_005265554.2 XP_005265611.1 15 2360
          XM_005265556.1 XP_005265613.1 14 1556
          XM_006713385.1 XP_006713448.1 27 2321
          XM_006713386.1 XP_006713449.1 26 2282
          XM_006713387.1 XP_006713450.1 26 2279
          XM_006713388.1 XP_006713451.1 26 2276
          XM_006713389.1 XP_006713452.1 26 2273
          XM_006713390.1 XP_006713453.1 27 2316
          XM_006713392.1 XP_006713455.1 26 2252
          XM_006713393.1 XP_006713456.1 26 2234
          XM_006713394.1 XP_006713457.1 25 2231
          XM_006713395.1 XP_006713458.1 26 2231
          XM_006713396.1 XP_006713459.1 25 2216
          XM_006713397.1 XP_006713460.1 24 2108
          XM_006713400.1 XP_006713463.1 16 1673
          XM_006713404.1 XP_006713467.1 13 1406
          XM_011534217.1 XP_011532519.1 27 2324
          XM_011534218.1 XP_011532520.1 28 2367
          XM_011534219.1 XP_011532521.1 28 2435
          XM_011534221.1 XP_011532523.1 25 2216
          XM_011534222.1 XP_011532524.1 25 2129
          XM_011534229.1 XP_011532531.1 17 1716
          XM_017007465.1 XP_016862954.1 24 2171
          XM_017007466.1 XP_016862955.1 25 2165
          XM_017007467.1 XP_016862956.1 25 2159
          XM_017007468.1 XP_016862957.1 22 2063
          XM_017007469.1 XP_016862958.1 24 2057
          XM_017007470.1 XP_016862959.1 22 2021
          XM_017007471.1 XP_016862960.1 17 1730
          XM_017007472.1 XP_016862961.1 17 1688
          XM_017007473.1 XP_016862962.1 16 1658
          XM_017007474.1 XP_016862963.1 16 1637
          XM_017007475.1 XP_016862964.1 16 2369
          XM_017007476.1 XP_016862965.1 16 1671
          XM_017007477.1 XP_016862966.1 15 2297
          XM_017007478.1 XP_016862967.1 14 1535
          XM_017007479.1 XP_016862968.1 15 1535
          XM_017007480.1 XP_016862969.1 14 1779
          XM_017007481.1 XP_016862970.1 14 1526
          XM_017007482.1 XP_016862971.1 13 1463
          XM_017007483.1 XP_016862972.1 13 2165
          XM_017007484.1 XP_016862973.1 14 1476
          GenBank mRNA AF115509.1 - 26 2015
          AK000255.1 - 11 1079
          AK024692.1 - 5 412
          AK124432.1 - 4 3190
          AK291514.1 - 13 1443
          AK302282.1 - 17 1429
          BC053668.1 - 12 1351 6146978
          BX648794.1 - 9 845
          CR749705.1 - 3 1783

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          biological_process
          Wnt signaling pathway
          protein binding
          LRR domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0