Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRRFIP2 SIRNA Assays ›
Gene Symbol:
LRRFIP2
Gene Name:
LRR binding FLII interacting protein 2
Gene Aliases:
HUFI-2
Chromosome Location:
Chr.3: 37052626 - 37176817 on Build GRCh38
UniGene ID:
Hs.740522
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
108005
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LRRFIP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134369.2
NP_001127841.1
15
1673
NM_001282691.1
NP_001269620.1
19
1662
NM_006309.3
NP_006300.1
28
2263
NM_017724.2
NP_060194.1
14
1601
XM_005265539.2
XP_005265596.1
29
3263
XM_005265540.1
XP_005265597.1
30
2546
XM_005265550.1
XP_005265607.1
21
2099
XM_005265551.1
XP_005265608.1
20
1988
XM_005265553.1
XP_005265610.1
17
1835
XM_005265554.2
XP_005265611.1
17
2567
XM_005265556.1
XP_005265613.1
16
1763
XM_006713385.1
XP_006713448.1
29
2528
XM_006713386.1
XP_006713449.1
28
2489
XM_006713387.1
XP_006713450.1
28
2486
XM_006713388.1
XP_006713451.1
28
2483
XM_006713389.1
XP_006713452.1
28
2480
XM_006713390.1
XP_006713453.1
29
2523
XM_006713392.1
XP_006713455.1
28
2459
XM_006713393.1
XP_006713456.1
28
2441
XM_006713394.1
XP_006713457.1
27
2438
XM_006713395.1
XP_006713458.1
28
2438
XM_006713396.1
XP_006713459.1
27
2423
XM_006713397.1
XP_006713460.1
26
2315
XM_006713400.1
XP_006713463.1
18
1880
XM_006713404.1
XP_006713467.1
15
1613
XM_011534217.1
XP_011532519.1
29
2531
XM_011534218.1
XP_011532520.1
30
2574
XM_011534219.1
XP_011532521.1
30
2642
XM_011534221.1
XP_011532523.1
27
2423
XM_011534222.1
XP_011532524.1
27
2336
XM_011534229.1
XP_011532531.1
19
1923
XM_017007465.1
XP_016862954.1
26
2378
XM_017007466.1
XP_016862955.1
27
2372
XM_017007467.1
XP_016862956.1
27
2366
XM_017007468.1
XP_016862957.1
24
2270
XM_017007469.1
XP_016862958.1
26
2264
XM_017007470.1
XP_016862959.1
24
2228
XM_017007471.1
XP_016862960.1
19
1937
XM_017007472.1
XP_016862961.1
19
1895
XM_017007473.1
XP_016862962.1
18
1865
XM_017007474.1
XP_016862963.1
18
1844
XM_017007475.1
XP_016862964.1
18
2576
XM_017007476.1
XP_016862965.1
18
1878
XM_017007477.1
XP_016862966.1
17
2504
XM_017007478.1
XP_016862967.1
16
1742
XM_017007479.1
XP_016862968.1
17
1742
XM_017007480.1
XP_016862969.1
16
1986
XM_017007481.1
XP_016862970.1
16
1733
XM_017007482.1
XP_016862971.1
15
1670
XM_017007483.1
XP_016862972.1
15
2372
XM_017007484.1
XP_016862973.1
16
1683
GenBank mRNA
AF115509.1
-
28
2222
AK000255.1
-
13
1286
AK024692.1
-
7
619
AK026336.1
-
2
102
AK124432.1
-
6
3397
AK291514.1
-
15
1650
AK302282.1
-
19
1636
BC053668.1
-
14
1558
6146978
BX648794.1
-
11
1052
CR749705.1
-
5
1990
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
biological_process
Wnt signaling pathway
protein binding
LRR domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0