Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NCOA2 SIRNA Assays ›
          Gene Symbol:
          NCOA2
          Gene Name:
          nuclear receptor coactivator 2
          Gene Aliases:
          GRIP1, KAT13C, NCoA-2, SRC2, TIF2, bHLHe75
          Chromosome Location:
          Chr.8: 70109762 - 70405390 on Build GRCh38
          UniGene ID:
          Hs.446678
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 108049

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NCOA2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321703.1 NP_001308632.1 9 1234
          NM_001321707.1 NP_001308636.1 9 1080
          NM_001321711.1 NP_001308640.1 9 1162
          NM_001321712.1 NP_001308641.1 9 1079
          NM_001321713.1 NP_001308642.1 8 975
          NM_006540.3 NP_006531.1 9 1162
          XM_017012961.1 XP_016868450.1 9 1149
          XM_017012962.1 XP_016868451.1 9 1173
          XM_017012963.1 XP_016868452.1 9 2386
          XM_017012964.1 XP_016868453.1 9 1441
          XM_017012965.1 XP_016868454.1 9 1106
          XM_017012966.1 XP_016868455.1 10 1370
          XM_017012967.1 XP_016868456.1 8 1084
          XM_017012968.1 XP_016868457.1 10 1171
          XM_017012969.1 XP_016868458.1 8 1202
          XM_017012970.1 XP_016868459.1 8 7621
          XM_017012971.1 XP_016868460.1 8 1333
          XM_017012972.1 XP_016868461.1 5 664
          XM_017012973.1 XP_016868462.1 9 1149
          GenBank mRNA AK308426.1 - 9 1127
          BC114383.1 - 8 1029 40082387
          X97674.1 - 9 1107

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          male gonad development
          regulation of glucose metabolic process
          bile acid and bile salt transport
          histone acetylation
          cerebellum development
          intracellular receptor signaling pathway
          cellular response to hormone stimulus
          circadian regulation of gene expression
          cellular lipid metabolic process
          locomotor rhythm
          positive regulation of female receptivity
          positive regulation of transcription from RNA polymerase II promoter
          cellular response to Thyroglobulin triiodothyronine
          positive regulation of receptor activity
          positive regulation of glucocorticoid receptor signaling pathway
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II intronic transcription regulatory region sequence-specific DNA binding
          chromatin binding
          transcription coactivator activity
          histone acetyltransferase activity
          protein binding
          ligand-dependent nuclear receptor binding
          PDZ domain binding
          estrogen receptor binding
          ligand-dependent nuclear receptor transcription coactivator activity
          thyroid hormone receptor coactivator activity
          protein complex binding
          progesterone receptor binding
          nuclear hormone receptor binding
          glucocorticoid receptor binding
          retinoid X receptor binding
          thyroid hormone receptor binding
          protein dimerization activity
          DNA polymerase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0