Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NCOA6 SIRNA Assays ›
Gene Symbol:
NCOA6
Gene Name:
nuclear receptor coactivator 6
Gene Aliases:
AIB3, ASC2, NRC, PRIP, RAP250, TRBP
Chromosome Location:
Chr.20: 34714774 - 34825649 on Build GRCh38
UniGene ID:
Hs.368971
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
108475
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NCOA6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242539.2
NP_001229468.1
5
1503
NM_001318240.1
NP_001305169.1
8
4025
NM_014071.4
NP_054790.2
7
1795
XM_006723755.3
XP_006723818.1
7
1723
XM_011528721.2
XP_011527023.1
7
1740
XM_011528726.2
XP_011527028.1
7
1740
XM_011528727.2
XP_011527029.1
7
1740
XM_017027739.1
XP_016883228.1
7
1740
XM_017027740.1
XP_016883229.1
8
1859
XM_017027741.1
XP_016883230.1
7
1740
XM_017027742.1
XP_016883231.1
7
1725
XM_017027743.1
XP_016883232.1
7
1740
XM_017027744.1
XP_016883233.1
7
1722
XM_017027745.1
XP_016883234.1
6
1596
XM_017027746.1
XP_016883235.1
7
1740
XM_017027747.1
XP_016883236.1
4
1117
XM_017027748.1
XP_016883237.1
7
1740
XM_017027749.1
XP_016883238.1
7
1740
XM_017027750.1
XP_016883239.1
7
1740
XM_017027751.1
XP_016883240.1
7
1723
GenBank mRNA
AF128458.1
-
6
1656
AF171667.1
-
6
1598
AF177388.1
-
6
1614
AF208227.1
-
8
4022
AF245115.1
-
5
1513
BC136272.1
-
6
1604
9020259
D80003.2
-
6
1591
JF707637.1
-
5
1454
JF707638.1
-
5
1454
JF707639.1
-
5
1454
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001276714, XM_008762295, XM_006235297, XM_008762296, XM_006235296, and others.
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
DNA replication
DNA repair
DNA recombination
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
cellular response to DNA damage stimulus
brain development
heart development
response to hormone
myeloid cell differentiation
intracellular estrogen receptor signaling pathway
positive regulation of insulin secretion involved in cellular response to glucose stimulus
glucocorticoid receptor signaling pathway
cellular lipid metabolic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
chromatin binding
transcription coactivator activity
protein binding
ligand-dependent nuclear receptor binding
enzyme binding
protein domain specific binding
estrogen receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
glucocorticoid receptor binding
protein homodimerization activity
peroxisome proliferator activated receptor binding
retinoid X receptor binding
thyroid hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0