Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST18 SIRNA Assays ›
Gene Symbol:
ST18
Gene Name:
ST18, C2H2C-type zinc finger
Gene Aliases:
NZF3, ZC2H2C3, ZC2HC10, ZNF387
Chromosome Location:
Chr.8: 52110832 - 52409879 on Build GRCh38
UniGene ID:
Hs.655499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
108614
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST18
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014682.2
NP_055497.1
10
1173
XM_006716487.1
XP_006716550.1
10
940
XM_011517629.1
XP_011515931.1
11
1096
XM_011517631.1
XP_011515933.1
12
1137
XM_011517632.1
XP_011515934.1
10
951
XM_011517633.1
XP_011515935.1
10
944
XM_011517634.1
XP_011515936.1
10
1140
XM_011517635.1
XP_011515937.1
10
1144
XM_011517636.2
XP_011515938.1
9
1221
XM_011517637.1
XP_011515939.1
8
678
XM_011517638.2
XP_011515940.1
7
719
XM_011517641.1
XP_011515943.1
12
1184
XM_011517642.1
XP_011515944.1
11
1096
XM_017014047.1
XP_016869536.1
10
1300
XM_017014048.1
XP_016869537.1
10
1304
XM_017014049.1
XP_016869538.1
11
1350
XM_017014050.1
XP_016869539.1
10
1137
XM_017014051.1
XP_016869540.1
9
1144
XM_017014052.1
XP_016869541.1
11
1354
XM_017014053.1
XP_016869542.1
11
1341
XM_017014054.1
XP_016869543.1
9
1390
XM_017014055.1
XP_016869544.1
9
981
XM_017014056.1
XP_016869545.1
12
1391
XM_017014057.1
XP_016869546.1
10
1194
XM_017014058.1
XP_016869547.1
10
955
XM_017014059.1
XP_016869548.1
10
1141
XM_017014060.1
XP_016869549.1
11
1178
XM_017014061.1
XP_016869550.1
9
1313
XM_017014062.1
XP_016869551.1
8
1065
XM_017014063.1
XP_016869552.1
9
1611
XM_017014064.1
XP_016869553.1
9
1225
XM_017014065.1
XP_016869554.1
11
1345
XM_017014066.1
XP_016869555.1
9
984
XM_017014067.1
XP_016869556.1
9
1394
XM_017014068.1
XP_016869557.1
10
1185
XM_017014069.1
XP_016869558.1
7
882
XM_017014070.1
XP_016869559.1
6
803
XM_017014071.1
XP_016869560.1
7
1051
XM_017014072.1
XP_016869561.1
10
1302
XM_017014073.1
XP_016869562.1
10
940
XM_017014074.1
XP_016869563.1
9
1223
XM_017014075.1
XP_016869564.1
11
1387
XM_017014076.1
XP_016869565.1
11
1391
XM_017014077.1
XP_016869566.1
11
1028
XM_017014078.1
XP_016869567.1
10
1308
XM_017014079.1
XP_016869568.1
10
1699
XM_017014080.1
XP_016869569.1
8
940
XM_017014081.1
XP_016869570.1
10
1307
XM_017014082.1
XP_016869571.1
10
1142
XM_017014083.1
XP_016869572.1
9
982
XM_017014084.1
XP_016869573.1
7
720
GenBank mRNA
AB011107.1
-
10
1037
BC031075.1
-
10
1070
BC117147.1
-
5
430
40125698
BC117149.1
-
5
430
40125700
BC143872.1
-
5
425
BC143873.1
-
6
518
Back To Top
More Information
Panther Classification:
Molecular Function -
zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0