Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMTF1 SIRNA Assays ›
Gene Symbol:
DMTF1
Gene Name:
cyclin D binding myb like transcription factor 1
Gene Aliases:
DMP1, DMTF, MRUL, hDMP1
Chromosome Location:
Chr.7: 87152361 - 87196332 on Build GRCh38
UniGene ID:
Hs.196129
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
109342
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMTF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142326.1
NP_001135798.1
14
1742
NM_001142327.1
NP_001135799.1
15
1859
NM_021145.3
NP_066968.3
17
2096
NR_024549.1
15
2070
NR_024550.1
15
2031
XM_011516735.2
XP_011515037.1
15
2781
XM_011516737.1
XP_011515039.1
16
1906
XM_011516739.2
XP_011515041.1
14
1671
XM_017012865.1
XP_016868354.1
16
2875
XM_017012866.1
XP_016868355.1
14
2658
XM_017012867.1
XP_016868356.1
13
3326
XM_017012868.1
XP_016868357.1
16
1906
XM_017012869.1
XP_016868358.1
17
1935
XM_017012870.1
XP_016868359.1
14
1674
XM_017012871.1
XP_016868360.1
16
3047
XM_017012872.1
XP_016868361.1
15
2953
XM_017012873.1
XP_016868362.1
16
2842
XM_017012874.1
XP_016868363.1
16
2078
XM_017012875.1
XP_016868364.1
16
2881
XM_017012876.1
XP_016868365.1
14
1821
XM_017012877.1
XP_016868366.1
15
1940
XM_017012878.1
XP_016868367.1
17
2975
GenBank mRNA
AB209230.1
-
14
1918
AF084530.1
-
15
1808
AF202144.1
-
15
1980
AF202145.1
-
15
2019
AK002073.1
-
3
318
AK095828.1
-
3
2428
AK123259.1
-
1
760
AK126664.1
-
15
2526
AK296211.1
-
14
1635
AK314622.1
-
15
1795
BC007418.2
-
6
808
BC007447.2
-
6
808
BC070064.1
-
17
2042
4373991
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell cycle
cell differentiation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor recruiting
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0