Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HIPK2 SIRNA Assays ›
          Gene Symbol:
          HIPK2
          Gene Name:
          homeodomain interacting protein kinase 2
          Gene Aliases:
          PRO0593
          Chromosome Location:
          Chr.7: 139561570 - 139777894 on Build GRCh38
          UniGene ID:
          Hs.731417
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 109492

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HIPK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001113239.2 NP_001106710.1 5, 6 1713
          NM_022740.4 NP_073577.3 5, 6 1713
          XM_006715935.2 XP_006715998.1 6, 7 1702
          XM_011516077.2 XP_011514379.1 5, 6 2291
          XM_011516078.2 XP_011514380.1 5, 6 2292
          XM_011516079.2 XP_011514381.1 5, 6 2291
          XM_011516080.2 XP_011514382.1 5, 6 2292
          XM_011516081.2 XP_011514383.1 5, 6 1688
          XM_017012069.1 XP_016867558.1 5, 6 1794
          GenBank mRNA AF207702.1 - 4, 5 1421
          AF208291.1 - 5, 6 1550
          AF326592.1 - 5, 6 1597

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          eye development
          transcription, DNA-templated
          protein phosphorylation
          DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
          transforming growth factor beta receptor signaling pathway
          smoothened signaling pathway
          adult walking behavior
          positive regulation of cell proliferation
          anterior/posterior pattern specification
          retina layer formation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          modulation by virus of host morphology or physiology
          neuron differentiation
          erythrocyte differentiation
          positive regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of BMP signaling pathway
          PML body organization
          positive regulation of protein binding
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation of DNA binding
          negative regulation of neuron apoptotic process
          positive regulation of angiogenesis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          embryonic camera-type eye morphogenesis
          voluntary musculoskeletal movement
          positive regulation of sequence-specific DNA binding transcription factor activity
          regulation of cell cycle
          embryonic retina morphogenesis in camera-type eye
          lens induction in camera-type eye
          SMAD protein signal transduction
          iris morphogenesis
          cellular response to hypoxia
          intrinsic apoptotic signaling pathway
          regulation of signal transduction by p53 class mediator
          RNA polymerase II activating transcription factor binding
          RNA polymerase II transcription coactivator activity
          transcription corepressor activity
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          SMAD binding
          virion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0