Search
Search

WDR24| Gene Symbol: WDR24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032259.3 | NP_115635.1 | 2 | 1394 | ||
| XM_011522699.2 | XP_011521001.1 | 3 | 1898 | |||
| GenBank mRNA | AK301146.1 | - | 2 | 1378 | ||
| AL136863.1 | - | 2 | 1371 | |||
| BC008025.1 | - | 2 | 859 | 3544791 | ||
| BC009761.2 | - | 2 | 394 | |||