Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FN1 SIRNA Assays ›
Gene Symbol:
FN1
Gene Name:
fibronectin 1
Gene Aliases:
CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF
Chromosome Location:
Chr.2: 215360440 - 215436167 on Build GRCh38
UniGene ID:
Hs.203717
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
11012
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306129.1
NP_001293058.1
6
986
NM_001306130.1
NP_001293059.1
6
986
NM_001306131.1
NP_001293060.1
6
986
NM_001306132.1
NP_001293061.1
6
986
NM_002026.3
NP_002017.1
6
984
NM_054034.2
NP_473375.2
6
984
NM_212474.2
NP_997639.1
6
984
NM_212476.2
NP_997641.1
6
984
NM_212478.2
NP_997643.1
6
984
NM_212482.2
NP_997647.1
6
984
XM_005246397.1
XP_005246454.1
6
1083
XM_005246398.1
XP_005246455.1
6
1083
XM_005246399.1
XP_005246456.1
6
1083
XM_005246401.1
XP_005246458.1
6
1083
XM_005246402.1
XP_005246459.1
6
1083
XM_005246403.1
XP_005246460.1
6
1083
XM_005246404.1
XP_005246461.1
6
1083
XM_005246405.1
XP_005246462.1
6
1083
XM_005246406.1
XP_005246463.1
6
1083
XM_005246407.1
XP_005246464.1
6
1083
XM_005246408.1
XP_005246465.1
6
1083
XM_005246409.1
XP_005246466.1
6
1083
XM_005246410.1
XP_005246467.1
6
1083
XM_005246411.1
XP_005246468.1
6
1083
XM_005246412.1
XP_005246469.1
6
1083
XM_005246414.1
XP_005246471.1
6
1083
XM_005246416.1
XP_005246473.1
6
1083
XM_017003692.1
XP_016859181.1
6
1083
XM_017003693.1
XP_016859182.1
6
1083
XM_017003694.1
XP_016859183.1
6
1083
XM_017003695.1
XP_016859184.1
6
1083
GenBank mRNA
AB191261.1
-
6
984
AB209840.1
-
6
976
AJ276395.1
-
6
774
AJ535086.1
-
6
774
AJ849445.1
-
6
774
AL832202.1
-
6
985
AL832771.1
-
6
985
BC117176.1
-
6
964
40125727
BC143754.1
-
6
964
9052268
BC143763.1
-
6
964
9052277
BX537590.1
-
6
985
BX538017.1
-
6
985
BX538018.1
-
6
985
BX538045.1
-
6
1024
BX640608.1
-
6
984
BX640731.1
-
6
985
BX640802.1
-
6
987
BX640803.1
-
6
984
BX640875.1
-
6
985
BX649182.1
-
6
987
CR749281.1
-
6
1023
CR749316.1
-
6
983
CR749317.1
-
6
983
U42404.1
-
4
413
X02761.1
-
6
638
Back To Top
More Information
Panther Classification:
Molecular Function -
extracellular matrix structural protein
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
cell activation
regulation of protein phosphorylation
platelet degranulation
response to ischemia
acute-phase response
cell-substrate junction assembly
cell adhesion
calcium-independent cell-matrix adhesion
positive regulation of cell proliferation
glial cell migration
regulation of cell shape
response to wounding
response to ozone
positive regulation of gene expression
positive regulation of peptidase activity
peptide cross-linking
extracellular matrix disassembly
extracellular matrix organization
integrin activation
substrate adhesion-dependent cell spreading
cellular response to vascular endothelial growth factor stimulus
endodermal cell differentiation
cellular response to platelet-derived growth factor stimulus
wound healing
negative regulation of apoptotic process
positive regulation of axon extension
positive regulation of fibroblast proliferation
leukocyte migration
positive regulation of chemotaxis
response to glucocorticoid
regulation of ERK1 and ERK2 cascade
cellular response to lipopolysaccharide
cellular response to mercury ion
cellular response to glucose stimulus
cellular response to interleukin-1
cellular response to prostaglandin E stimulus
cellular response to transforming growth factor beta stimulus
cellular response to BMP stimulus
positive regulation of substrate-dependent cell migration, cell attachment to substrate
negative regulation of transforming growth factor-beta secretion
protease binding
integrin binding
protein binding
collagen binding
heparin binding
peptidase activator activity
identical protein binding
mercury ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0