Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KMT2C SIRNA Assays ›
          Gene Symbol:
          KMT2C LOC105378260
          Gene Name:
          lysine methyltransferase 2C
          histone-lysine N-methyltransferase 2C
          Gene Aliases:
          HALR, MLL3
          -
          Chromosome Location:
          Chr.7: 152134925 - 152436642 on Build GRCh38
          Chromosome Location:
          Chr.21: 10462898 - 10514209 on Build GRCh38
          UniGene ID:
          Hs.647120
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 110283

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KMT2C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_170606.2 NP_733751.2 23, 24 3942
          XM_005250025.4 XP_005250082.1 24, 25 3748
          XM_005250026.3 XP_005250083.1 24, 25 3782
          XM_005250027.4 XP_005250084.1 23, 24 3956
          XM_005250028.4 XP_005250085.1 23, 24 3956
          XM_005250031.4 XP_005250088.1 23, 24 3956
          XM_006716077.3 XP_006716140.1 23, 24 3956
          XM_006716078.3 XP_006716141.1 23, 24 3956
          XM_006716079.3 XP_006716142.1 23, 24 3956
          XM_011516450.2 XP_011514752.1 23, 24 3956
          XM_011516451.2 XP_011514753.1 22, 23 3836
          XM_011516452.2 XP_011514754.1 23, 24 3956
          XM_011516453.2 XP_011514755.1 23, 24 3956
          XM_011516454.2 XP_011514756.1 18, 19 2951
          XM_011516456.2 XP_011514758.1 23, 24 3956
          XM_017012480.1 XP_016867969.1 24, 25 3785
          XM_017012481.1 XP_016867970.1 24, 25 3745
          XM_017012482.1 XP_016867971.1 23, 24 3956
          XM_017012483.1 XP_016867972.1 23, 24 3956
          XM_017012484.1 XP_016867973.1 23, 24 3923
          XM_017012485.1 XP_016867974.1 23, 24 3953
          XM_017012486.1 XP_016867975.1 23, 24 3956
          XM_017012487.1 XP_016867976.1 23, 24 3924
          XM_017012488.1 XP_016867977.1 23, 24 3956
          XM_017012489.1 XP_016867978.1 4, 5 463
          XM_017012490.1 XP_016867979.1 1, 2 35
          GenBank mRNA AB040939.2 - 12, 13 2058
          AF264750.2 - 9, 10 1269
          AK095628.1 - 16, 17 2664
          AY024361.1 - 23, 24 3903
          BC151837.1 - 12, 13 2058
          Gene Symbol: LOC105378260
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017028520.1 XP_016884009.1 7, 8 1382

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: XM_008762670.
          This siRNA also targets the following transcripts in Mouse: NM_001081383, AY036886, AK087368, AY138582, BC037044.

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          histone H3-K4 methylation
          DNA binding
          protein binding
          zinc ion binding
          transferase activity, transferring acyl groups
          histone-lysine N-methyltransferase activity
          histone methyltransferase activity (H3-K4 specific)
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0