Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KMT2C SIRNA Assays ›
Gene Symbol:
KMT2C
Gene Name:
lysine methyltransferase 2C
Gene Aliases:
HALR, MLL3
Chromosome Location:
Chr.7: 152134925 - 152436642 on Build GRCh38
UniGene ID:
Hs.647120
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
110284
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KMT2C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_170606.2
NP_733751.2
24
4023
XM_005250025.4
XP_005250082.1
25
3829
XM_005250026.3
XP_005250083.1
25
3863
XM_005250027.4
XP_005250084.1
24
4037
XM_005250028.4
XP_005250085.1
24
4037
XM_005250031.4
XP_005250088.1
24
4037
XM_006716077.3
XP_006716140.1
24
4037
XM_006716078.3
XP_006716141.1
24
4037
XM_006716079.3
XP_006716142.1
24
4037
XM_011516450.2
XP_011514752.1
24
4037
XM_011516451.2
XP_011514753.1
23
3917
XM_011516452.2
XP_011514754.1
24
4037
XM_011516453.2
XP_011514755.1
24
4037
XM_011516454.2
XP_011514756.1
19
3032
XM_011516456.2
XP_011514758.1
24
4037
XM_017012480.1
XP_016867969.1
25
3866
XM_017012481.1
XP_016867970.1
25
3826
XM_017012482.1
XP_016867971.1
24
4037
XM_017012483.1
XP_016867972.1
24
4037
XM_017012484.1
XP_016867973.1
24
4004
XM_017012485.1
XP_016867974.1
24
4034
XM_017012486.1
XP_016867975.1
24
4037
XM_017012487.1
XP_016867976.1
24
4005
XM_017012488.1
XP_016867977.1
24
4037
XM_017012489.1
XP_016867978.1
5
544
XM_017012490.1
XP_016867979.1
2
116
GenBank mRNA
AB040939.2
-
13
2139
AF264750.2
-
10
1350
AK025911.1
-
14
2246
AY024361.1
-
24
3984
BC151837.1
-
13
2139
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
histone H3-K4 methylation
DNA binding
protein binding
zinc ion binding
transferase activity, transferring acyl groups
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K4 specific)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0