Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HIPK1 SIRNA Assays ›
          Gene Symbol:
          HIPK1
          Gene Name:
          homeodomain interacting protein kinase 1
          Gene Aliases:
          Myak, Nbak2
          Chromosome Location:
          Chr.1: 113929197 - 113977869 on Build GRCh38
          UniGene ID:
          Hs.532363
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 111083

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HIPK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_152696.3 NP_689909.2 9 2193
          NM_181358.2 NP_852003.1 7 913
          NM_198268.2 NP_938009.1 9 2193
          NM_198269.2 NP_938010.1 8 1104
          XM_005270609.3 XP_005270666.1 9 2370
          XM_005270610.4 XP_005270667.1 9 2370
          XM_005270611.4 XP_005270668.1 9 2370
          XM_005270612.4 XP_005270669.1 9 2332
          XM_005270613.4 XP_005270670.1 11 2283
          XM_006710443.3 XP_006710506.1 9 2268
          XM_011540977.2 XP_011539279.1 8 2250
          XM_017000605.1 XP_016856094.1 9 2332
          GenBank mRNA AB089957.1 - 9 2157
          AK122662.1 - 9 2191
          AK302726.1 - 9 2377
          AL833829.1 - 8 1082
          BC028408.1 - 8 1109 4821915
          BC033012.2 - 9 2193 4830408
          BC036057.1 - 7 913 5272816

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          eye development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein phosphorylation
          smoothened signaling pathway
          positive regulation of cell proliferation
          anterior/posterior pattern specification
          regulation of tumor necrosis factor-mediated signaling pathway
          retina layer formation
          neuron differentiation
          adherens junction assembly
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation of angiogenesis
          embryonic camera-type eye morphogenesis
          embryonic retina morphogenesis in camera-type eye
          definitive hemopoiesis
          lens induction in camera-type eye
          iris morphogenesis
          endothelial cell apoptotic process
          extrinsic apoptotic signaling pathway
          regulation of signal transduction by p53 class mediator
          protein serine/threonine kinase activity
          protein binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline