Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TXNRD1 SIRNA Assays ›
          Gene Symbol:
          TXNRD1
          Gene Name:
          thioredoxin reductase 1
          Gene Aliases:
          GRIM-12, TR, TR1, TRXR1, TXNR
          Chromosome Location:
          Chr.12: 104215779 - 104350307 on Build GRCh38
          UniGene ID:
          Hs.654922
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 111300

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TXNRD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001093771.2 NP_001087240.1 8, 9 898
          NM_001261445.1 NP_001248374.1 6, 7 1320
          NM_001261446.1 NP_001248375.1 5, 6 875
          NM_003330.3 NP_003321.3 6, 7 1235
          NM_182729.2 NP_877393.1 6, 7 998
          NM_182742.2 NP_877419.1 5, 6 1125
          NM_182743.2 NP_877420.1 5, 6 888
          GenBank mRNA AF077367.1 - 5, 6 552
          AF208018.1 - 4, 5 448
          AJ001050.1 - 4, 5 461
          AK128515.1 - 5, 6 783
          AK226071.1 - 6, 7 706
          AK293322.1 - 5, 6 535
          AK295067.1 - 6, 7 634
          AK296495.1 - 5, 6 525
          AK304241.1 - 8, 9 881
          AK308881.1 - 6, 7 929
          AK312262.1 - 6, 7 897
          AY057105.1 - 8, 9 874
          BC018122.1 - 6, 7 611 3883490
          BT019640.1 - 4, 5 424
          CR536506.1 - 4, 5 424
          D88687.1 - 6, 7 676
          S79851.1 - 6, 7 863
          X91247.1 - 6, 7 863

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          mesoderm formation
          selenium compound metabolic process
          signal transduction
          cell proliferation
          nucleobase-containing small molecule interconversion
          cell redox homeostasis
          oxidation-reduction process
          cellular oxidant detoxification
          thioredoxin-disulfide reductase activity
          protein binding
          electron carrier activity
          protein disulfide oxidoreductase activity
          flavin adenine dinucleotide binding
          methylselenol reductase activity
          methylseleninic acid reductase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0