Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003358.2 | NP_003349.1 | 1, 2 | 556 | ||
XM_005252186.4 | XP_005252243.1 | 1, 2 | 501 | |||
XM_017015107.1 | XP_016870596.1 | 1, 2 | 215 | |||
GenBank mRNA | AK314847.1 | - | 1, 2 | 161 | ||
BC038711.1 | - | 1, 2 | 557 | 5295561 | ||
D50840.1 | - | 1, 2 | 396 |