Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ASPH SIRNA Assays ›
Gene Symbol:
ASPH
Gene Name:
aspartate beta-hydroxylase
Gene Aliases:
AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin
Chromosome Location:
Chr.8: 61500556 - 61714640 on Build GRCh38
UniGene ID:
Hs.332422
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
111447
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ASPH
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164750.1
NP_001158222.1
2
249
NM_001164751.1
NP_001158223.1
2
249
NM_001164752.1
NP_001158224.1
2
249
NM_001164753.1
NP_001158225.1
2
249
NM_001164754.1
NP_001158226.1
2
444
NM_001164755.1
NP_001158227.1
2
444
NM_001164756.1
NP_001158228.1
2
444
NM_004318.3
NP_004309.2
2
444
NM_020164.4
NP_064549.1
2
249
NM_032466.3
NP_115855.1
2
444
NM_032467.3
NP_115856.1
2
249
NM_032468.4
NP_115857.1
2
249
XM_005251235.2
XP_005251292.1
2
413
XM_005251236.2
XP_005251293.1
2
413
XM_005251238.2
XP_005251295.1
2
413
XM_005251239.2
XP_005251296.1
2
413
XM_005251240.1
XP_005251297.1
2
191
XM_005251242.2
XP_005251299.1
2
413
XM_005251243.2
XP_005251300.1
2
413
XM_005251244.1
XP_005251301.1
2
191
XM_005251246.2
XP_005251303.1
2
413
XM_005251247.2
XP_005251304.1
2
413
XM_005251248.1
XP_005251305.1
2
99
XM_005251250.2
XP_005251307.1
2
413
XM_017013419.1
XP_016868908.1
2
413
XM_017013420.1
XP_016868909.1
2
413
XM_017013421.1
XP_016868910.1
2
413
XM_017013422.1
XP_016868911.1
2
413
XM_017013423.1
XP_016868912.1
2
413
XM_017013424.1
XP_016868913.1
2
413
XM_017013425.1
XP_016868914.1
2
413
XM_017013426.1
XP_016868915.1
2
191
XM_017013427.1
XP_016868916.1
2
191
XM_017013428.1
XP_016868917.1
2
413
XM_017013429.1
XP_016868918.1
2
413
XM_017013430.1
XP_016868919.1
2
413
XM_017013431.1
XP_016868920.1
2
99
XM_017013432.1
XP_016868921.1
2
413
XM_017013433.1
XP_016868922.1
2
99
XM_017013434.1
XP_016868923.1
2
413
XM_017013435.1
XP_016868924.1
2
191
XM_017013436.1
XP_016868925.1
2
413
XM_017013437.1
XP_016868926.1
2
413
XM_017013438.1
XP_016868927.1
2
413
XM_017013439.1
XP_016868928.1
2
99
XM_017013440.1
XP_016868929.1
2
191
XM_017013441.1
XP_016868930.1
2
413
XM_017013442.1
XP_016868931.1
2
413
XM_017013443.1
XP_016868932.1
2
413
XM_017013444.1
XP_016868933.1
2
99
XM_017013445.1
XP_016868934.1
2
99
XM_017013446.1
XP_016868935.1
2
99
XM_017013447.1
XP_016868936.1
2
99
GenBank mRNA
AF184241.1
-
2
129
AF224468.1
-
2
199
AF224469.1
-
2
174
AF289489.1
-
2
186
AF306765.1
-
2
170
AK291836.1
-
2
257
AK295528.1
-
2
213
AK298577.1
-
3
365
AK304314.1
-
2
205
BC015518.2
-
2
363
BC025236.1
-
2
372
BC066929.1
-
2
340
BC142967.1
-
2
321
BC144362.1
-
2
179
BC144363.1
-
2
180
CR996516.1
-
2
444
DA136318.1
-
2
413
DN993915.1
-
2
257
EU718494.1
-
2
252
FJ461473.1
-
2
175
S83325.1
-
2
186
U03109.1
-
2
252
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
pattern specification process
negative regulation of cell proliferation
regulation of protein stability
ion transmembrane transport
limb morphogenesis
peptidyl-aspartic acid hydroxylation
positive regulation of proteolysis
oxidation-reduction process
palate development
face morphogenesis
activation of cysteine-type endopeptidase activity
regulation of protein depolymerization
regulation of cardiac conduction
peptide-aspartate beta-dioxygenase activity
structural molecule activity
calcium ion binding
protein binding
structural constituent of muscle
electron carrier activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0