Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HSPD1 SIRNA Assays ›
          Gene Symbol:
          HSPD1
          Gene Name:
          heat shock protein family D (Hsp60) member 1
          Gene Aliases:
          CPN60, GROEL, HLD4, HSP-60, HSP60, HSP65, HuCHA60, SPG13
          Chromosome Location:
          Chr.2: 197486584 - 197500274 on Build GRCh38
          UniGene ID:
          Hs.595053
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 11163

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HSPD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002156.4 NP_002147.2 2, 3 319
          NM_199440.1 NP_955472.1 2, 3 299
          GenBank mRNA AK298368.1 - 2, 3 238
          AK298632.1 - 2, 3 269
          AK298844.1 - 2, 3 238
          AK301276.1 - 2, 3 274
          AK312240.1 - 2, 3 252
          BC002676.2 - 2, 3 229 3609039
          BC003030.1 - 2, 3 228 2821157
          BC047350.1 - 1, 2 389
          BC067082.1 - 2, 3 227 4025725
          BC073746.1 - 2, 3 227 6372068
          BG701476.1 - 2, 3 320
          BX647145.1 - 2, 3 307
          EU719626.1 - 1, 2 182
          KU178139.1 - 1, 2 182
          KU178141.1 - 1, 2 182
          M22382.1 - 2, 3 227
          M34664.1 - 2, 3 206

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_022229, XM_006244932, X53585, BC086507, X54793.
          This siRNA also targets the following transcripts in Mouse: X53584.

          Gene Ontology Categories:

          Function(s) Process(es)

          B cell cytokine production
          MyD88-dependent toll-like receptor signaling pathway
          positive regulation of T cell mediated immune response to tumor cell
          'de novo' protein folding
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          response to unfolded protein
          response to cold
          viral process
          positive regulation of interferon-alpha production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-6 production
          protein refolding
          B cell proliferation
          T cell activation
          B cell activation
          positive regulation of macrophage activation
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          isotype switching to IgG isotypes
          protein stabilization
          positive regulation of T cell activation
          chaperone-mediated protein complex assembly
          protein maturation
          lipopolysaccharide binding
          p53 binding
          DNA replication origin binding
          single-stranded DNA binding
          double-stranded RNA binding
          protein binding
          ATP binding
          ATPase activity
          ubiquitin protein ligase binding
          poly(A) RNA binding
          unfolded protein binding
          chaperone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0