Search
Search

ALG12| Gene Symbol: ALG12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024105.3 | NP_077010.1 | 2, 3 | 441 | ||
| XM_011530371.2 | XP_011528673.1 | 2, 3 | 452 | |||
| XM_017028936.1 | XP_016884425.1 | 2, 3 | 452 | |||
| XM_017028937.1 | XP_016884426.1 | 2, 3 | 452 | |||
| GenBank mRNA | AF318343.1 | - | 2, 3 | 393 | ||
| AJ290427.1 | - | 2, 3 | 314 | |||
| AJ303120.1 | - | 2, 3 | 314 | |||
| BC001729.1 | - | 2, 3 | 288 | 3353085 | ||
| BC098562.1 | - | 2, 3 | 334 | 30560020 | ||
| CB858085.1 | - | 2, 3 | 442 | |||
| CR456369.1 | - | 1, 2 | 230 | |||