Search
Search

GALNT14| Gene Symbol: GALNT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253826.1 | NP_001240755.1 | 7 | 1241 | ||
| NM_001253827.1 | NP_001240756.1 | 8 | 1006 | |||
| NM_024572.3 | NP_078848.2 | 6 | 1226 | |||
| XM_011533105.1 | XP_011531407.1 | 8 | 760 | |||
| XM_017004906.1 | XP_016860395.1 | 6 | 691 | |||
| XM_017004907.1 | XP_016860396.1 | 6 | 702 | |||
| XM_017004908.1 | XP_016860397.1 | 6 | 985 | |||
| GenBank mRNA | AB078144.1 | - | 6 | 586 | ||
| AK022753.1 | - | 7 | 662 | |||
| AK091313.1 | - | 6 | 596 | |||
| AK122747.1 | - | 8 | 990 | |||
| AK225808.1 | - | 6 | 950 | |||
| AK298735.1 | - | 7 | 654 | |||
| AK309417.1 | - | 8 | 845 | |||
| AY358758.1 | - | 6 | 919 | |||
| BC010659.1 | - | 6 | 1226 | 3874588 | ||
| Y09324.4 | - | 6 | 586 | |||