Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KCNMA1 SIRNA Assays ›
          Gene Symbol:
          KCNMA1
          Gene Name:
          potassium calcium-activated channel subfamily M alpha 1
          Gene Aliases:
          BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
          Chromosome Location:
          Chr.10: 76869601 - 77637819 on Build GRCh38
          UniGene ID:
          Hs.144795
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 112880

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KCNMA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001014797.2 NP_001014797.1 3, 4 788
          NM_001161352.1 NP_001154824.1 3, 4 788
          NM_001161353.1 NP_001154825.1 3, 4 788
          NM_001271518.1 NP_001258447.1 2, 3 626
          NM_001271519.1 NP_001258448.1 3, 4 788
          NM_001322829.1 NP_001309758.1 3, 4 788
          NM_001322830.1 NP_001309759.1 3, 4 788
          NM_001322832.1 NP_001309761.1 3, 4 788
          NM_001322835.1 NP_001309764.1 3, 4 788
          NM_001322836.1 NP_001309765.1 3, 4 788
          NM_001322837.1 NP_001309766.1 3, 4 788
          NM_002247.3 NP_002238.2 3, 4 788
          XM_005269776.3 XP_005269833.1 3, 4 777
          XM_005269778.1 XP_005269835.1 3, 4 777
          XM_005269781.2 XP_005269838.1 3, 4 709
          XM_005269787.3 XP_005269844.1 3, 4 777
          XM_005269789.2 XP_005269846.1 3, 4 709
          XM_005269792.2 XP_005269849.1 3, 4 709
          XM_005269796.2 XP_005269853.1 3, 4 709
          XM_006717826.2 XP_006717889.1 3, 4 709
          XM_011539773.1 XP_011538075.1 4, 5 909
          XM_011539774.1 XP_011538076.1 4, 5 909
          XM_011539775.1 XP_011538077.1 4, 5 909
          XM_011539776.1 XP_011538078.1 4, 5 909
          XM_011539777.1 XP_011538079.1 3, 4 777
          XM_011539778.2 XP_011538080.1 3, 4 709
          XM_011539779.1 XP_011538081.1 4, 5 909
          XM_011539780.1 XP_011538082.1 3, 4 777
          XM_011539781.2 XP_011538083.1 4, 5 909
          XM_011539782.2 XP_011538084.1 4, 5 861
          XM_011539783.1 XP_011538085.1 3, 4 777
          XM_011539784.1 XP_011538086.1 4, 5 909
          XM_011539785.1 XP_011538087.1 3, 4 777
          XM_017016207.1 XP_016871696.1 4, 5 909
          XM_017016208.1 XP_016871697.1 4, 5 909
          XM_017016209.1 XP_016871698.1 4, 5 909
          XM_017016210.1 XP_016871699.1 4, 5 909
          XM_017016211.1 XP_016871700.1 4, 5 909
          XM_017016212.1 XP_016871701.1 4, 5 909
          XM_017016213.1 XP_016871702.1 4, 5 909
          XM_017016214.1 XP_016871703.1 4, 5 909
          XM_017016215.1 XP_016871704.1 3, 4 777
          XM_017016216.1 XP_016871705.1 4, 5 909
          XM_017016217.1 XP_016871706.1 4, 5 861
          XM_017016218.1 XP_016871707.1 2, 3 234
          XM_017016219.1 XP_016871708.1 4, 5 909
          XM_017016222.1 XP_016871711.1 3, 4 777
          GenBank mRNA AB113382.1 - 3, 4 553
          AB113575.1 - 3, 4 539
          AB209488.1 - 2, 3 194
          AB524033.1 - 2, 3 449
          AF025999.1 - 3, 4 539
          AF118141.1 - 3, 4 416
          AY040849.1 - 3, 4 553
          BC137115.1 - 3, 4 691 9021112
          BC137137.1 - 3, 4 691 9021134
          BC144496.1 - 3, 4 691 9053023
          U09384.1 - 3, 4 606
          U11058.2 - 3, 4 502
          U11717.1 - 3, 4 563
          U13913.1 - 3, 4 689
          U23767.1 - 3, 4 611

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031828, AY344964, AY330292, AF135265, U40603, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001253365, NM_001253366, NM_001253378, NM_001253377, NM_001253367, and others.

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          potassium ion transport
          response to osmotic stress
          cellular potassium ion homeostasis
          response to carbon monoxide
          regulation of ion transmembrane transport
          regulation of membrane potential
          positive regulation of apoptotic process
          negative regulation of cell volume
          response to calcium ion
          micturition
          smooth muscle contraction involved in micturition
          potassium ion transmembrane transport
          actin binding
          voltage-gated potassium channel activity
          protein binding
          calcium-activated potassium channel activity
          metal ion binding
          large conductance calcium-activated potassium channel activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0