Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KCNMA1 SIRNA Assays ›
Gene Symbol:
KCNMA1
Gene Name:
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases:
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
Chromosome Location:
Chr.10: 76869601 - 77637819 on Build GRCh38
UniGene ID:
Hs.144795
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
112882
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KCNMA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001014797.2
NP_001014797.1
25, 26
3172
NM_001161352.1
NP_001154824.1
25, 26
3334
NM_001161353.1
NP_001154825.1
25, 26
3283
NM_001271518.1
NP_001258447.1
24, 25
3010
NM_001271519.1
NP_001258448.1
26, 27
3250
NM_001322829.1
NP_001309758.1
25, 26
3169
NM_001322830.1
NP_001309759.1
27, 28
3262
NM_001322832.1
NP_001309761.1
24, 25
3160
NM_001322835.1
NP_001309764.1
26, 27
3253
NM_001322836.1
NP_001309765.1
25, 26
3169
NM_001322837.1
NP_001309766.1
26, 27
3253
NM_001322838.1
NP_001309767.1
26, 27
3207
NM_002247.3
NP_002238.2
24, 25
3160
XM_005269776.3
XP_005269833.1
25, 26
3230
XM_005269778.1
XP_005269835.1
26, 27
3335
XM_005269781.2
XP_005269838.1
25, 26
3255
XM_005269787.3
XP_005269844.1
24, 25
3149
XM_005269789.2
XP_005269846.1
26, 27
3174
XM_005269792.2
XP_005269849.1
25, 26
3168
XM_005269796.2
XP_005269853.1
24, 25
3081
XM_006717826.2
XP_006717889.1
25, 26
3162
XM_011539773.1
XP_011538075.1
27, 28
3467
XM_011539774.1
XP_011538076.1
28, 29
3461
XM_011539775.1
XP_011538077.1
26, 27
3455
XM_011539776.1
XP_011538078.1
26, 27
3455
XM_011539777.1
XP_011538079.1
27, 28
3416
XM_011539778.2
XP_011538080.1
26, 27
3336
XM_011539779.1
XP_011538081.1
26, 27
3368
XM_011539780.1
XP_011538082.1
26, 27
3335
XM_011539781.2
XP_011538083.1
26, 27
3293
XM_011539782.2
XP_011538084.1
25, 26
3233
XM_011539783.1
XP_011538085.1
26, 27
3248
XM_011539785.1
XP_011538087.1
26, 27
3239
XM_017016207.1
XP_016871696.1
28, 29
3548
XM_017016208.1
XP_016871697.1
28, 29
3548
XM_017016209.1
XP_016871698.1
27, 28
3536
XM_017016210.1
XP_016871699.1
27, 28
3467
XM_017016211.1
XP_016871700.1
27, 28
3380
XM_017016212.1
XP_016871701.1
27, 28
3374
XM_017016213.1
XP_016871702.1
26, 27
3368
XM_017016214.1
XP_016871703.1
26, 27
3362
XM_017016215.1
XP_016871704.1
27, 28
3329
XM_017016216.1
XP_016871705.1
26, 27
3293
XM_017016217.1
XP_016871706.1
25, 26
3233
XM_017016218.1
XP_016871707.1
25, 26
2699
XM_017016222.1
XP_016871711.1
24, 25
3149
XM_017016223.1
XP_016871712.1
23, 24
2545
GenBank mRNA
AB113382.1
-
25, 26
3099
AB113575.1
-
25, 26
3034
AB209488.1
-
24, 25
2579
AB524033.1
-
24, 25
2833
AF025999.1
-
24, 25
2911
AF118141.1
-
24, 25
2788
AK310379.1
-
26, 27
3207
AY040849.1
-
24, 25
2925
AY445624.1
-
3, 4
279
BC137115.1
-
24, 25
3063
9021112
BC137137.1
-
24, 25
3063
9021134
BC144496.1
-
26, 27
3153
9053023
BX648925.1
-
21, 22
2325
U02632.1
-
18, 19
2105
U11058.2
-
24, 25
2874
U11717.1
-
24, 25
2935
U13913.1
-
24, 25
3061
U23767.1
-
24, 25
2983
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: HQ221756.
Panther Classification:
Molecular Function -
ion channel
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
potassium ion transport
response to osmotic stress
cellular potassium ion homeostasis
response to carbon monoxide
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of apoptotic process
negative regulation of cell volume
response to calcium ion
micturition
smooth muscle contraction involved in micturition
potassium ion transmembrane transport
actin binding
voltage-gated potassium channel activity
protein binding
calcium-activated potassium channel activity
metal ion binding
large conductance calcium-activated potassium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0