Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CTSH SIRNA Assays ›
          Gene Symbol:
          CTSH
          Gene Name:
          cathepsin H
          Gene Aliases:
          ACC-4, ACC-5, ACC4, ACC5, CPSB
          Chromosome Location:
          Chr.15: 78921750 - 78945098 on Build GRCh38
          UniGene ID:
          Hs.148641
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 113112

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CTSH
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001319137.1 NP_001306066.1 9 1358
          NM_004390.4 NP_004381.2 8 685
          XM_005254181.2 XP_005254238.1 8 789
          XM_017021951.1 XP_016877440.1 9 771
          XM_017021952.1 XP_016877441.1 8 985
          GenBank mRNA AF426247.1 - 8 568
          AF426248.1 - 7 532
          AK130158.1 - 8 634
          AK309491.1 - 9 1306
          AK314698.1 - 8 633
          BC002479.2 - 8 629 3349910
          BC018821.2 - 8 629
          BG775606.1 - 8 665
          CR456881.1 - 8 568
          X07549.1 - 5 309
          X16832.1 - 8 602
          Y18461.1 - 3 157

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          metanephros development
          T cell mediated cytotoxicity
          adaptive immune response
          immune response-regulating signaling pathway
          proteolysis
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of cell proliferation
          positive regulation of gene expression
          positive regulation of epithelial cell migration
          neuropeptide catabolic process
          bradykinin catabolic process
          positive regulation of peptidase activity
          antigen processing and presentation
          positive regulation of cell migration
          zymogen activation
          protein destabilization
          response to retinoic acid
          membrane protein proteolysis
          negative regulation of apoptotic process
          surfactant homeostasis
          cellular protein metabolic process
          positive regulation of angiogenesis
          proteolysis involved in cellular protein catabolic process
          dichotomous subdivision of terminal units involved in lung branching
          ERK1 and ERK2 cascade
          cellular response to thyroid hormone stimulus
          positive regulation of apoptotic signaling pathway
          endopeptidase activity
          aminopeptidase activity
          cysteine-type endopeptidase activity
          serine-type endopeptidase activity
          protein binding
          peptidase activity
          cysteine-type peptidase activity
          cysteine-type endopeptidase activator activity involved in apoptotic process
          HLA-A specific activating MHC class I receptor activity
          thyroid hormone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0