Search
Search

AGBL1| Gene Symbol: AGBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011521226.2 | XP_011519528.1 | 11 | 1873 | ||
| XM_011521227.2 | XP_011519529.1 | 11 | 1873 | |||
| XM_011521228.2 | XP_011519530.1 | 11 | 1873 | |||
| XM_011521230.2 | XP_011519532.1 | 11 | 1873 | |||
| XM_017021918.1 | XP_016877407.1 | 11 | 1840 | |||
| XM_017021919.1 | XP_016877408.1 | 11 | 1873 | |||
| XM_017021920.1 | XP_016877409.1 | 11 | 1873 | |||
| XM_017021921.1 | XP_016877410.1 | 11 | 1873 | |||
| GenBank mRNA | AK056872.1 | - | 3 | 609 | ||
| BC128152.1 | - | 2 | 719 | |||