Search
Search

CHRNB4| Gene Symbol: CHRNB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000750.3 | NP_000741.1 | 2 | 233 | ||
| NM_001256567.1 | NP_001243496.1 | 2 | 233 | |||
| XM_011521186.2 | XP_011519488.1 | 4 | 263 | |||
| XM_011521187.2 | XP_011519489.1 | 3 | 170 | |||
| XM_011521192.2 | XP_011519494.1 | 2 | 226 | |||
| XM_017021885.1 | XP_016877374.1 | 2 | 328 | |||
| XM_017021886.1 | XP_016877375.1 | 2 | 2135 | |||
| XM_017021887.1 | XP_016877376.1 | 2 | 137 | |||
| XM_017021888.1 | XP_016877377.1 | 2 | 137 | |||
| XM_017021889.1 | XP_016877378.1 | 2 | 137 | |||
| GenBank mRNA | AK310443.1 | - | 8 | 983 | ||
| BC096080.1 | - | 2 | 236 | 40000733 | ||
| BC096081.1 | - | 2 | 236 | |||
| BC096082.1 | - | 2 | 236 | 40000737 | ||
| BC096083.1 | - | 2 | 236 | |||
| U48861.1 | - | 2 | 241 | |||
| U62439.1 | - | 2 | 207 | |||
| Y08416.1 | - | 2 | 212 | |||