Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HDAC9 SIRNA Assays ›
Gene Symbol:
HDAC9
Gene Name:
histone deacetylase 9
Gene Aliases:
HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
Chromosome Location:
Chr.7: 18086942 - 19003745 on Build GRCh38
UniGene ID:
Hs.196054
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115115
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HDAC9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204144.2
NP_001191073.1
5
715
NM_001204145.2
NP_001191074.1
3
407
NM_001204146.2
NP_001191075.1
3
576
NM_001204147.2
NP_001191076.1
2
373
NM_001204148.2
NP_001191077.1
2
373
NM_001321868.1
NP_001308797.1
4
655
NM_001321869.1
NP_001308798.1
4
655
NM_001321870.1
NP_001308799.1
4
655
NM_001321871.1
NP_001308800.1
4
655
NM_001321872.1
NP_001308801.1
4
655
NM_001321873.1
NP_001308802.1
4
655
NM_001321874.1
NP_001308803.1
3
576
NM_001321875.1
NP_001308804.1
3
576
NM_001321876.1
NP_001308805.1
3
576
NM_001321877.1
NP_001308806.1
3
576
NM_001321878.1
NP_001308807.1
3
576
NM_001321879.1
NP_001308808.1
3
576
NM_001321884.1
NP_001308813.1
3
576
NM_001321885.1
NP_001308814.1
3
576
NM_001321886.1
NP_001308815.1
4
635
NM_001321887.1
NP_001308816.1
4
466
NM_001321888.1
NP_001308817.1
3
407
NM_001321889.1
NP_001308818.1
4
635
NM_001321890.1
NP_001308819.1
3
576
NM_001321891.1
NP_001308820.1
3
576
NM_001321893.1
NP_001308822.1
4
473
NM_001321894.1
NP_001308823.1
4
473
NM_001321895.1
NP_001308824.1
3
407
NM_001321896.1
NP_001308825.1
4
745
NM_001321897.1
NP_001308826.1
4
473
NM_001321898.1
NP_001308827.1
3
407
NM_001321899.1
NP_001308828.1
4
473
NM_001321900.1
NP_001308829.1
3
576
NM_001321901.1
NP_001308830.1
3
407
NM_001321902.1
NP_001308831.1
3
407
NM_014707.3
NP_055522.1
3
407
NM_058176.2
NP_478056.1
2
262
NM_178423.2
NP_848510.1
3
407
NM_178425.3
NP_848512.1
2
262
XM_011515629.2
XP_011513931.1
3
356
XM_011515631.2
XP_011513933.1
4
655
XM_011515634.2
XP_011513936.1
3
356
XM_011515635.2
XP_011513937.1
4
1207
XM_011515636.2
XP_011513938.1
3
937
XM_011515637.2
XP_011513939.1
3
361
XM_011515638.2
XP_011513940.1
3
937
XM_011515639.2
XP_011513941.1
3
1134
XM_011515641.2
XP_011513943.1
4
1204
XM_011515642.2
XP_011513944.1
2
1933
XM_011515644.2
XP_011513946.1
4
1194
XM_011515645.2
XP_011513947.1
2
1933
XM_011515646.2
XP_011513948.1
2
1933
XM_011515648.2
XP_011513950.1
3
937
XM_011515649.2
XP_011513951.1
3
1141
XM_011515651.1
XP_011513953.1
3
337
XM_017012826.1
XP_016868315.1
3
937
XM_017012827.1
XP_016868316.1
4
996
XM_017012828.1
XP_016868317.1
3
1136
XM_017012829.1
XP_016868318.1
3
337
XM_017012830.1
XP_016868319.1
3
337
XM_017012831.1
XP_016868320.1
3
337
XM_017012832.1
XP_016868321.1
3
337
GenBank mRNA
AB018287.1
-
3
371
AJ459808.1
-
3
371
AK097178.1
-
3
356
AK296214.1
-
3
308
AK297404.1
-
3
361
AK304298.1
-
5
708
AK304343.1
-
2
293
AK304410.1
-
2
336
AK316109.1
-
3
527
AY032737.1
-
2
221
AY032738.1
-
2
221
AY197371.1
-
2
221
AY429540.1
-
3
461
BC111735.1
-
2
220
BC150328.1
-
3
382
8860495
BC152405.1
-
3
371
8862508
CR749550.1
-
2
329
DC369193.1
-
2
369
DC369899.1
-
2
373
Back To Top
More Information
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to amphetamine
transcription, DNA-templated
inflammatory response
heart development
chromatin modification
histone deacetylation
neuron differentiation
B cell differentiation
cellular response to insulin stimulus
peptidyl-lysine deacetylation
B cell activation
negative regulation of transcription, DNA-templated
regulation of skeletal muscle fiber development
regulation of striated muscle cell differentiation
histone H3 deacetylation
histone H4 deacetylation
positive regulation of cell migration involved in sprouting angiogenesis
transcription corepressor activity
histone deacetylase activity
protein kinase C binding
protein binding
transcription factor binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
histone deacetylase binding
metal ion binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0