Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HDAC9 SIRNA Assays ›
Gene Symbol:
HDAC9
Gene Name:
histone deacetylase 9
Gene Aliases:
HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
Chromosome Location:
Chr.7: 18086942 - 19003745 on Build GRCh38
UniGene ID:
Hs.196054
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115116
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HDAC9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204144.2
NP_001191073.1
10
1292
NM_001204145.2
NP_001191074.1
8
984
NM_001204146.2
NP_001191075.1
8
1162
NM_001204147.2
NP_001191076.1
7
944
NM_001204148.2
NP_001191077.1
8
1091
NM_001321868.1
NP_001308797.1
9
1232
NM_001321869.1
NP_001308798.1
10
1373
NM_001321870.1
NP_001308799.1
10
1364
NM_001321871.1
NP_001308800.1
9
1247
NM_001321872.1
NP_001308801.1
9
1241
NM_001321873.1
NP_001308802.1
9
1235
NM_001321874.1
NP_001308803.1
9
1294
NM_001321875.1
NP_001308804.1
9
1300
NM_001321876.1
NP_001308805.1
9
1288
NM_001321877.1
NP_001308806.1
8
1162
NM_001321878.1
NP_001308807.1
9
1285
NM_001321879.1
NP_001308808.1
8
1156
NM_001321884.1
NP_001308813.1
9
1288
NM_001321885.1
NP_001308814.1
8
1177
NM_001321886.1
NP_001308815.1
10
1344
NM_001321887.1
NP_001308816.1
10
1178
NM_001321888.1
NP_001308817.1
8
978
NM_001321889.1
NP_001308818.1
9
1215
NM_001321890.1
NP_001308819.1
9
1279
NM_001321891.1
NP_001308820.1
8
1153
NM_001321893.1
NP_001308822.1
9
1050
NM_001321894.1
NP_001308823.1
9
1059
NM_001321895.1
NP_001308824.1
8
993
NM_001321896.1
NP_001308825.1
10
1463
NM_001321897.1
NP_001308826.1
9
1059
NM_001321898.1
NP_001308827.1
8
1008
NM_001321899.1
NP_001308828.1
10
1191
NM_001321900.1
NP_001308829.1
9
1285
NM_001321901.1
NP_001308830.1
9
1119
NM_001321902.1
NP_001308831.1
9
1125
NM_014707.3
NP_055522.1
9
1116
NM_058176.2
NP_478056.1
8
971
NM_178423.2
NP_848510.1
9
1116
NM_178425.3
NP_848512.1
8
980
XM_011515629.2
XP_011513931.1
9
1074
XM_011515631.2
XP_011513933.1
10
1388
XM_011515634.2
XP_011513936.1
8
942
XM_011515635.2
XP_011513937.1
10
1940
XM_011515636.2
XP_011513938.1
9
1661
XM_011515637.2
XP_011513939.1
9
1085
XM_011515638.2
XP_011513940.1
9
1655
XM_011515639.2
XP_011513941.1
9
1852
XM_011515641.2
XP_011513943.1
10
1937
XM_011515642.2
XP_011513944.1
8
2666
XM_011515644.2
XP_011513946.1
10
1912
XM_011515645.2
XP_011513947.1
8
2651
XM_011515646.2
XP_011513948.1
8
2642
XM_011515648.2
XP_011513950.1
9
1670
XM_011515649.2
XP_011513951.1
9
1874
XM_011515651.1
XP_011513953.1
8
908
XM_017012826.1
XP_016868315.1
9
1646
XM_017012827.1
XP_016868316.1
10
1729
XM_017012828.1
XP_016868317.1
8
1722
XM_017012829.1
XP_016868318.1
9
1049
XM_017012830.1
XP_016868319.1
9
1040
XM_017012831.1
XP_016868320.1
8
917
XM_017012832.1
XP_016868321.1
9
1049
GenBank mRNA
AB018287.1
-
9
1080
AF124924.1
-
6
638
AJ459808.1
-
9
1080
AK097178.1
-
8
942
AK296214.1
-
8
894
AK297404.1
-
8
938
AK304298.1
-
10
1285
AK304410.1
-
7
907
AK316109.1
-
8
1113
AY032737.1
-
8
930
AY032738.1
-
8
930
AY197371.1
-
8
939
BC111735.1
-
7
806
BC150328.1
-
9
1091
8860495
BC152405.1
-
9
1080
8862508
CR749550.1
-
8
1047
Back To Top
More Information
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to amphetamine
transcription, DNA-templated
inflammatory response
heart development
chromatin modification
histone deacetylation
neuron differentiation
B cell differentiation
cellular response to insulin stimulus
peptidyl-lysine deacetylation
B cell activation
negative regulation of transcription, DNA-templated
regulation of skeletal muscle fiber development
regulation of striated muscle cell differentiation
histone H3 deacetylation
histone H4 deacetylation
positive regulation of cell migration involved in sprouting angiogenesis
transcription corepressor activity
histone deacetylase activity
protein kinase C binding
protein binding
transcription factor binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
histone deacetylase binding
metal ion binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0