Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCL6 SIRNA Assays ›
          Gene Symbol:
          BCL6
          Gene Name:
          B-cell CLL/lymphoma 6
          Gene Aliases:
          BCL5, BCL6A, LAZ3, ZBTB27, ZNF51
          Chromosome Location:
          Chr.3: 187721377 - 187745725 on Build GRCh38
          UniGene ID:
          Hs.478588
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 115226

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BCL6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130845.1 NP_001124317.1 4 669
          NM_001134738.1 NP_001128210.1 3 261
          NM_001706.4 NP_001697.2 4 577
          XM_005247694.3 XP_005247751.1 4 3921
          XM_011513062.2 XP_011511364.1 4 577
          GenBank mRNA AK304325.1 - 4 320
          AK308162.1 - 4 319
          BC142705.1 - 4 292 40148809
          BC150184.1 - 4 293 40148297
          EU139066.1 - 2 236
          EU883531.1 - 2 212
          S67779.1 - 4 631
          U00115.1 - 4 539
          Z21943.1 - 4 296

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus, translocation
          negative regulation of transcription from RNA polymerase II promoter
          cell morphogenesis
          negative regulation of cell-matrix adhesion
          germinal center formation
          regulation of germinal center formation
          negative regulation of B cell apoptotic process
          transcription, DNA-templated
          inflammatory response
          cellular response to DNA damage stimulus
          Rho protein signal transduction
          spermatogenesis
          negative regulation of cell proliferation
          actin cytoskeleton organization
          myeloid cell differentiation
          B cell differentiation
          negative regulation of cell growth
          positive regulation of B cell proliferation
          positive regulation of histone deacetylation
          negative regulation of mast cell cytokine production
          negative regulation of Rho protein signal transduction
          type 2 immune response
          regulation of cell proliferation
          regulation of apoptotic process
          positive regulation of apoptotic process
          regulation of GTPase activity
          regulation of memory T cell differentiation
          negative regulation of T-helper 2 cell differentiation
          positive regulation of neuron differentiation
          negative regulation of Notch signaling pathway
          negative regulation of transcription, DNA-templated
          negative regulation of isotype switching to IgE isotypes
          erythrocyte development
          regulation of inflammatory response
          regulation of neurogenesis
          regulation of immune response
          positive regulation of cellular component movement
          negative regulation of cellular senescence
          RNA polymerase II regulatory region sequence-specific DNA binding
          intronic transcription regulatory region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          chromatin DNA binding
          sequence-specific DNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0