Search
Search

COMMD3-BMI1
BMI1| Gene Symbol: COMMD3-BMI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204062.1 | NP_001190991.1 | 14 | 1374 | ||
| GenBank mRNA | AK313235.1 | - | 14 | 1349 | ||
| Gene Symbol: BMI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005180.8 | NP_005171.4 | 10 | 1416 | ||
| GenBank mRNA | BC011652.2 | - | 10 | 1415 | 4138748 | |
| BT019450.1 | - | 9 | 910 | |||
| CR456911.1 | - | 9 | 910 | |||
| L13689.1 | - | 10 | 1389 | |||