Search
Search

BMI1
COMMD3-BMI1| Gene Symbol: BMI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005180.8 | NP_005171.4 | 10 | 2507 | ||
| GenBank mRNA | BC011652.2 | - | 10 | 2506 | 4138748 | |
| BE008203.1 | - | 1 | 215 | |||
| L13689.1 | - | 10 | 2481 | |||
| Gene Symbol: COMMD3-BMI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204062.1 | NP_001190991.1 | 14 | 2465 | ||