Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MECOM SIRNA Assays ›
Gene Symbol:
MECOM
Gene Name:
MDS1 and EVI1 complex locus
Gene Aliases:
AML1-EVI-1, EVI1, MDS1, MDS1-EVI1, PRDM3, RUSAT2
Chromosome Location:
Chr.3: 169083499 - 169663781 on Build GRCh38
UniGene ID:
Hs.744090
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115627
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MECOM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105077.3
NP_001098547.3
5
420
NM_001105078.3
NP_001098548.2
4
414
NM_001163999.1
NP_001157471.1
4
358
NM_001164000.1
NP_001157472.1
4
1290
NM_001205194.1
NP_001192123.1
4
718
NM_004991.3
NP_004982.2
5
1056
NM_005241.3
NP_005232.2
4
358
XM_005247213.3
XP_005247270.1
5
1062
XM_005247214.3
XP_005247271.1
5
1062
XM_005247215.3
XP_005247272.1
5
1061
XM_005247219.2
XP_005247276.1
4
1027
XM_005247220.2
XP_005247277.1
4
368
XM_005247221.2
XP_005247278.1
4
351
XM_005247223.2
XP_005247280.1
4
1027
XM_005247224.3
XP_005247281.1
5
1062
XM_005247225.3
XP_005247282.1
5
1061
XM_005247226.3
XP_005247283.1
5
939
XM_011512546.2
XP_011510848.1
4
815
XM_017005874.1
XP_016861363.1
4
1730
XM_017005875.1
XP_016861364.1
4
368
XM_017005876.1
XP_016861365.1
4
1027
XM_017005877.1
XP_016861366.1
5
1061
XM_017005878.1
XP_016861367.1
4
1027
GenBank mRNA
AF164154.1
-
4
1559
AF164155.1
-
5
1314
AF164156.1
-
5
875
AF164157.1
-
5
402
AK292865.1
-
4
341
AK304098.1
-
5
1002
AK308404.1
-
5
701
AK310509.1
-
3
202
BC130520.1
-
5
394
40146551
BC143951.1
-
5
394
9052469
BC143952.1
-
4
332
9052470
BX640908.1
-
4
1290
BX647613.1
-
4
1022
GQ352634.1
-
4
622
X54989.1
-
4
356
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
apoptotic process
cell differentiation
histone lysine methylation
negative regulation of programmed cell death
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
negative regulation of JNK cascade
regulation of cell cycle
hematopoietic stem cell proliferation
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
histone-lysine N-methyltransferase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0