Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRRFIP2 SIRNA Assays ›
Gene Symbol:
LRRFIP2
Gene Name:
LRR binding FLII interacting protein 2
Gene Aliases:
HUFI-2
Chromosome Location:
Chr.3: 37052626 - 37176817 on Build GRCh38
UniGene ID:
Hs.740522
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115787
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LRRFIP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134369.2
NP_001127841.1
13
1448
NM_001282691.1
NP_001269620.1
17
1437
NM_006309.3
NP_006300.1
26
2038
NM_017724.2
NP_060194.1
12
1376
XM_005265539.2
XP_005265596.1
27
3038
XM_005265540.1
XP_005265597.1
28
2321
XM_005265550.1
XP_005265607.1
19
1874
XM_005265551.1
XP_005265608.1
18
1763
XM_005265553.1
XP_005265610.1
15
1610
XM_005265554.2
XP_005265611.1
15
2342
XM_005265556.1
XP_005265613.1
14
1538
XM_006713385.1
XP_006713448.1
27
2303
XM_006713386.1
XP_006713449.1
26
2264
XM_006713387.1
XP_006713450.1
26
2261
XM_006713388.1
XP_006713451.1
26
2258
XM_006713389.1
XP_006713452.1
26
2255
XM_006713390.1
XP_006713453.1
27
2298
XM_006713392.1
XP_006713455.1
26
2234
XM_006713393.1
XP_006713456.1
26
2216
XM_006713394.1
XP_006713457.1
25
2213
XM_006713395.1
XP_006713458.1
26
2213
XM_006713396.1
XP_006713459.1
25
2198
XM_006713397.1
XP_006713460.1
24
2090
XM_006713400.1
XP_006713463.1
16
1655
XM_006713404.1
XP_006713467.1
13
1388
XM_011534217.1
XP_011532519.1
27
2306
XM_011534218.1
XP_011532520.1
28
2349
XM_011534219.1
XP_011532521.1
28
2417
XM_011534221.1
XP_011532523.1
25
2198
XM_011534222.1
XP_011532524.1
25
2111
XM_011534229.1
XP_011532531.1
17
1698
XM_017007465.1
XP_016862954.1
24
2153
XM_017007466.1
XP_016862955.1
25
2147
XM_017007467.1
XP_016862956.1
25
2141
XM_017007468.1
XP_016862957.1
22
2045
XM_017007469.1
XP_016862958.1
24
2039
XM_017007470.1
XP_016862959.1
22
2003
XM_017007471.1
XP_016862960.1
17
1712
XM_017007472.1
XP_016862961.1
17
1670
XM_017007473.1
XP_016862962.1
16
1640
XM_017007474.1
XP_016862963.1
16
1619
XM_017007475.1
XP_016862964.1
16
2351
XM_017007476.1
XP_016862965.1
16
1653
XM_017007477.1
XP_016862966.1
15
2279
XM_017007478.1
XP_016862967.1
14
1517
XM_017007479.1
XP_016862968.1
15
1517
XM_017007480.1
XP_016862969.1
14
1761
XM_017007481.1
XP_016862970.1
14
1508
XM_017007482.1
XP_016862971.1
13
1445
XM_017007483.1
XP_016862972.1
13
2147
XM_017007484.1
XP_016862973.1
14
1458
GenBank mRNA
AF115509.1
-
26
1997
AK000255.1
-
11
1061
AK024692.1
-
5
394
AK124432.1
-
4
3172
AK291514.1
-
13
1425
AK302282.1
-
17
1411
BC053668.1
-
12
1333
6146978
BX648794.1
-
9
827
CR749705.1
-
3
1765
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
biological_process
Wnt signaling pathway
protein binding
LRR domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0