Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRIM32 SIRNA Assays ›
          Gene Symbol:
          TRIM32
          Gene Name:
          tripartite motif containing 32
          Gene Aliases:
          BBS11, HT2A, LGMD2H, TATIP
          Chromosome Location:
          Chr.9: 116687302 - 116701300 on Build GRCh38
          UniGene ID:
          Hs.591910
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 115914

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TRIM32
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099679.1 NP_001093149.1 2 391
          NM_012210.3 NP_036342.2 2 394
          XM_005251813.3 XP_005251870.1 2 440
          XM_011518398.2 XP_011516700.1 2 428
          XM_017014486.1 XP_016869975.1 3 451
          GenBank mRNA AK314264.1 - 2 375
          AX775779.1 - 2 363
          BC003154.1 - 2 363 2906024
          BI544919.1 - 2 394
          DN991209.1 - 2 356
          U18543.1 - 2 343

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          tissue homeostasis
          actin ubiquitination
          response to UV
          protein ubiquitination
          positive regulation of cell growth
          positive regulation of cell migration
          regulation of type I interferon production
          negative regulation of viral transcription
          response to tumor necrosis factor
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          innate immune response
          fat cell differentiation
          positive regulation of neuron differentiation
          positive regulation of protein catabolic process
          positive regulation of cell cycle
          positive regulation of proteolysis
          muscle cell cellular homeostasis
          negative regulation of fibroblast proliferation
          positive regulation of neurogenesis
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of striated muscle cell differentiation
          axon development
          negative regulation of viral release from host cell
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          positive regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of interleukin-17-mediated signaling pathway
          positive regulation of chemokine (C-C motif) ligand 20 production
          positive regulation of cell motility
          transcription coactivator activity
          RNA binding
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          ligase activity
          myosin binding
          Tat protein binding
          translation initiation factor binding
          ubiquitin binding
          protein self-association
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0