Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST18 SIRNA Assays ›
Gene Symbol:
ST18
Gene Name:
ST18, C2H2C-type zinc finger
Gene Aliases:
NZF3, ZC2H2C3, ZC2HC10, ZNF387
Chromosome Location:
Chr.8: 52110832 - 52409879 on Build GRCh38
UniGene ID:
Hs.655499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
116038
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST18
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014682.2
NP_055497.1
8, 9
909
XM_006716487.1
XP_006716550.1
8, 9
676
XM_011517629.1
XP_011515931.1
9, 10
832
XM_011517631.1
XP_011515933.1
10, 11
873
XM_011517632.1
XP_011515934.1
8, 9
687
XM_011517633.1
XP_011515935.1
8, 9
680
XM_011517634.1
XP_011515936.1
8, 9
876
XM_011517635.1
XP_011515937.1
8, 9
880
XM_011517636.2
XP_011515938.1
7, 8
957
XM_011517637.1
XP_011515939.1
6, 7
414
XM_011517638.2
XP_011515940.1
5, 6
455
XM_011517642.1
XP_011515944.1
9, 10
832
XM_017014047.1
XP_016869536.1
8, 9
1036
XM_017014048.1
XP_016869537.1
8, 9
1040
XM_017014049.1
XP_016869538.1
9, 10
1086
XM_017014050.1
XP_016869539.1
8, 9
873
XM_017014051.1
XP_016869540.1
7, 8
880
XM_017014052.1
XP_016869541.1
9, 10
1090
XM_017014053.1
XP_016869542.1
9, 10
1077
XM_017014054.1
XP_016869543.1
7, 8
1126
XM_017014055.1
XP_016869544.1
7, 8
717
XM_017014056.1
XP_016869545.1
10, 11
1127
XM_017014057.1
XP_016869546.1
8, 9
930
XM_017014058.1
XP_016869547.1
8, 9
691
XM_017014059.1
XP_016869548.1
8, 9
877
XM_017014060.1
XP_016869549.1
9, 10
914
XM_017014061.1
XP_016869550.1
7, 8
1049
XM_017014062.1
XP_016869551.1
6, 7
801
XM_017014063.1
XP_016869552.1
7, 8
1347
XM_017014064.1
XP_016869553.1
7, 8
961
XM_017014065.1
XP_016869554.1
9, 10
1081
XM_017014066.1
XP_016869555.1
7, 8
720
XM_017014067.1
XP_016869556.1
7, 8
1130
XM_017014068.1
XP_016869557.1
8, 9
921
XM_017014069.1
XP_016869558.1
5, 6
618
XM_017014070.1
XP_016869559.1
4, 5
539
XM_017014071.1
XP_016869560.1
5, 6
787
XM_017014072.1
XP_016869561.1
8, 9
1038
XM_017014073.1
XP_016869562.1
8, 9
676
XM_017014074.1
XP_016869563.1
7, 8
959
XM_017014081.1
XP_016869570.1
8, 9
1043
XM_017014082.1
XP_016869571.1
8, 9
878
XM_017014083.1
XP_016869572.1
7, 8
718
XM_017014084.1
XP_016869573.1
5, 6
456
GenBank mRNA
AB011107.1
-
8, 9
773
BC031075.1
-
8, 9
806
BC117147.1
-
3, 4
166
40125698
BC117149.1
-
3, 4
166
40125700
BC143872.1
-
3, 4
165
Back To Top
More Information
Panther Classification:
Molecular Function -
zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0