Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SOD1 SIRNA Assays ›
Gene Symbol:
SOD1
Gene Name:
superoxide dismutase 1, soluble
Gene Aliases:
ALS, ALS1, HEL-S-44, IPOA, SOD, hSod1, homodimer
Chromosome Location:
Chr.21: 31659622 - 31668931 on Build GRCh38
UniGene ID:
Hs.443914
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
116854
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SOD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000454.4
NP_000445.1
5
922
GenBank mRNA
X02317.1
-
5
838
Back To Top
More Information
Panther Classification:
Molecular Function -
oxidoreductase
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
response to reactive oxygen species
response to superoxide
ovarian follicle development
positive regulation of cytokine production
placenta development
retina homeostasis
response to amphetamine
myeloid cell homeostasis
platelet degranulation
glutathione metabolic process
superoxide metabolic process
cellular iron ion homeostasis
spermatogenesis
embryo implantation
cell aging
sensory perception of sound
locomotory behavior
anterograde axonal transport
retrograde axonal transport
regulation of blood pressure
response to heat
response to organic substance
transmission of nerve impulse
removal of superoxide radicals
response to nutrient levels
peripheral nervous system myelin maintenance
positive regulation of superoxide anion generation
regulation of T cell differentiation in thymus
response to carbon monoxide
cellular response to potassium ion
regulation of multicellular organism growth
response to drug
response to hydrogen peroxide
superoxide anion generation
positive regulation of apoptotic process
positive regulation of catalytic activity
regulation of GTPase activity
negative regulation of neuron apoptotic process
response to ethanol
negative regulation of cholesterol biosynthetic process
regulation of protein kinase activity
regulation of organ growth
response to antibiotic
response to copper ion
muscle cell cellular homeostasis
thymus development
response to axon injury
hydrogen peroxide biosynthetic process
regulation of mitochondrial membrane potential
oxidation-reduction process
heart contraction
neurofilament cytoskeleton organization
relaxation of vascular smooth muscle
auditory receptor cell stereocilium organization
cellular response to cadmium ion
cellular response to ATP
reactive oxygen species metabolic process
response to antipsychotic drug
positive regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
superoxide dismutase activity
copper ion binding
protein binding
zinc ion binding
protein phosphatase 2B binding
identical protein binding
protein homodimerization activity
Rac GTPase binding
chaperone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-s6kwc:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0